missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ MQuant™ Peroxide Test Strips
To check for presence or absence of residues of Peroxide
Supplier: MilliporeSigma™ 1103370001
Includes: Tube containing 100 strips
Description
- Small and easy to handle
- Flexible and universal application
- Enzymatic reaction method ideal for screening
- Immediate result produced through on-site analysis
- PET film and low reagent content
- Easily disposable
1.10011.0001
Color-scale graduation: 0.5 - 2 - 5 - 10 - 25mg/L H2O2
1.10081.0001
Color-scale graduation: 1 - 3 - 10 - 30 - 100mg/L H2O2
1.10337.0001
Color-scale graduation: 100 - 200 - 400 - 600 - 800 - 1000mg/L H2O2
Reclose the tube containing the test strips immediately after use.
For food and water analysis
Specifications
For testing residual concentrations of disinfectants on equipment or the product itself | |
100 - 200 - 400 - 600 - 800 - 1000 | |
100 Strips/Pk. | |
2°C to 8°C | |
Peroxide Test Strips |
Colorimeter | |
100 | |
100 to 1000mg/L H2O2 | |
Enzymatic Reaction |