Learn More
N-(3-Aminopropyl)imidazole, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 351940500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N-(3-Aminopropyl)imidazole | |
5036-48-6 | |
100.0 | |
Colorless | |
296.0°C | |
Authentic | |
Glass bottle | |
1.5175 to 1.5195 | |
50 g | |
1-3-aminopropyl imidazole, n-3-aminopropyl imidazole, 1h-imidazole-1-propanamine, n-3-aminopropyl-imidazole, 3-1h-imidazol-1-yl propan-1-amine, 3-imidazol-1-ylpropylamine, 3-1h-imidazol-1-yl-1-propanamine, 1-3-aminopropyl-imidazole, 3-imidazol-1-yl propylamine, 3-imidazolylpropylamine | |
KDHWOCLBMVSZPG-UHFFFAOYSA-N | |
3-imidazol-1-ylpropan-1-amine | |
78736 | |
98% |
98% | |
97.5 | |
-68.0°C | |
1.0620g/mL | |
154°C | |
97.5% min. (GC) | |
C6H11N3 | |
MFCD00009819 | |
1.062 | |
Solubility in water: miscible. | |
NCCCN1C=CN=C1 | |
125.18 | |
125.17 | |
Liquid |
Chemical Identifiers
5036-48-6 | |
125.18 | |
KDHWOCLBMVSZPG-UHFFFAOYSA-N | |
78736 | |
NCCCN1C=CN=C1 |
C6H11N3 | |
MFCD00009819 | |
1-3-aminopropyl imidazole, n-3-aminopropyl imidazole, 1h-imidazole-1-propanamine, n-3-aminopropyl-imidazole, 3-1h-imidazol-1-yl propan-1-amine, 3-imidazol-1-ylpropylamine, 3-1h-imidazol-1-yl-1-propanamine, 1-3-aminopropyl-imidazole, 3-imidazol-1-yl propylamine, 3-imidazolylpropylamine | |
3-imidazol-1-ylpropan-1-amine |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful to aquatic life.
Harmful to aquatic life with long lasting effects.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED
GHS Signal Word: Danger
EINECSNumber : 225-730-9
TSCA : TSCA
RUO – Research Use Only