missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(3-Aminopropyl)imidazole, 98%, Thermo Scientific Chemicals

$90.26 - $290.57

Chemical Identifiers

CAS 5036-48-6
Molecular Formula C6H11N3
Molecular Weight (g/mol) 125.18
MDL Number MFCD00009819
InChI Key KDHWOCLBMVSZPG-UHFFFAOYSA-N
Synonym 1-3-aminopropyl imidazole, n-3-aminopropyl imidazole, 1h-imidazole-1-propanamine, n-3-aminopropyl-imidazole, 3-1h-imidazol-1-yl propan-1-amine, 3-imidazol-1-ylpropylamine, 3-1h-imidazol-1-yl-1-propanamine, 1-3-aminopropyl-imidazole, 3-imidazol-1-yl propylamine, 3-imidazolylpropylamine
PubChem CID 78736
IUPAC Name 3-imidazol-1-ylpropan-1-amine
SMILES NCCCN1C=CN=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC351940500
SDS View Documents
Thermo Scientific Chemicals
351940500
50 g Glass bottle
Each for $90.26
 
AC351942500
SDS View Documents
Thermo Scientific Chemicals
351942500
250 g Glass bottle
Each for $290.57
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5036-48-6
125.18
KDHWOCLBMVSZPG-UHFFFAOYSA-N
78736
NCCCN1C=CN=C1
C6H11N3
MFCD00009819
1-3-aminopropyl imidazole, n-3-aminopropyl imidazole, 1h-imidazole-1-propanamine, n-3-aminopropyl-imidazole, 3-1h-imidazol-1-yl propan-1-amine, 3-imidazol-1-ylpropylamine, 3-1h-imidazol-1-yl-1-propanamine, 1-3-aminopropyl-imidazole, 3-imidazol-1-yl propylamine, 3-imidazolylpropylamine
3-imidazol-1-ylpropan-1-amine

Specifications

5036-48-6
100.0
Colorless
296.0°C
Authentic
Glass bottle
1.5175 to 1.5195
50 g
1-3-aminopropyl imidazole, n-3-aminopropyl imidazole, 1h-imidazole-1-propanamine, n-3-aminopropyl-imidazole, 3-1h-imidazol-1-yl propan-1-amine, 3-imidazol-1-ylpropylamine, 3-1h-imidazol-1-yl-1-propanamine, 1-3-aminopropyl-imidazole, 3-imidazol-1-yl propylamine, 3-imidazolylpropylamine
KDHWOCLBMVSZPG-UHFFFAOYSA-N
3-imidazol-1-ylpropan-1-amine
78736
98%
N-(3-Aminopropyl)imidazole, 98%
97.5
-68.0°C
1.0620g/mL
154°C
97.5% min. (GC)
C6H11N3
MFCD00009819
1.062
Solubility in water: miscible.
NCCCN1C=CN=C1
125.18
125.17
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful to aquatic life.
Harmful to aquatic life with long lasting effects.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED

GHS Signal Word: Danger

EINECSNumber : 225-730-9

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only