missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(3-Fluorophenyl)thiourea, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals L0953803

 View more versions of this product

Catalog No. AAL0953803


Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

N-(3-Fluorophenyl)thiourea
114°C to 117°C
C7H7FN2S
1 g
2718454
YKOBLHQALWQKTC-UHFFFAOYSA-N
(3-fluorophenyl)thiourea
2734799
99%
458-05-9
Odorless
MFCD00041167
UN2811
1-3-fluorophenyl-2-thiourea, 1-3-fluorophenyl thiourea, 3-fluorophenyl thiourea, n-3-fluorophenyl thiourea, amino 3-fluorophenyl amino methane-1-thione, acmc-20apbn, 1-m-fluorophenyl thiourea, 3-fluoro-phenyl-thiourea
C1=CC(=CC(=C1)F)NC(=S)N
170.205
170.21

Chemical Identifiers

458-05-9
170.205
YKOBLHQALWQKTC-UHFFFAOYSA-N
2734799
C1=CC(=CC(=C1)F)NC(=S)N
C7H7FN2S
MFCD00041167
1-3-fluorophenyl-2-thiourea, 1-3-fluorophenyl thiourea, 3-fluorophenyl thiourea, n-3-fluorophenyl thiourea, amino 3-fluorophenyl amino methane-1-thione, acmc-20apbn, 1-m-fluorophenyl thiourea, 3-fluoro-phenyl-thiourea
(3-fluorophenyl)thiourea
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H301-H315-H319

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions