missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-(3-Fluorophenyl)thiourea, 99%, Thermo Scientific™
$55.12 - $55.12
Chemical Identifiers
CAS | 458-05-9 |
---|---|
Molecular Formula | C7H7FN2S |
Molecular Weight (g/mol) | 170.205 |
MDL Number | MFCD00041167 |
InChI Key | YKOBLHQALWQKTC-UHFFFAOYSA-N |
Synonym | 1-3-fluorophenyl-2-thiourea, 1-3-fluorophenyl thiourea, 3-fluorophenyl thiourea, n-3-fluorophenyl thiourea, amino 3-fluorophenyl amino methane-1-thione, acmc-20apbn, 1-m-fluorophenyl thiourea, 3-fluoro-phenyl-thiourea |
PubChem CID | 2734799 |
IUPAC Name | (3-fluorophenyl)thiourea |
SMILES | C1=CC(=CC(=C1)F)NC(=S)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0953803
|
Thermo Scientific Chemicals
L0953803 |
1 g |
N/A
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
458-05-9 | |
170.205 | |
YKOBLHQALWQKTC-UHFFFAOYSA-N | |
2734799 | |
C1=CC(=CC(=C1)F)NC(=S)N |
C7H7FN2S | |
MFCD00041167 | |
1-3-fluorophenyl-2-thiourea, 1-3-fluorophenyl thiourea, 3-fluorophenyl thiourea, n-3-fluorophenyl thiourea, amino 3-fluorophenyl amino methane-1-thione, acmc-20apbn, 1-m-fluorophenyl thiourea, 3-fluoro-phenyl-thiourea | |
(3-fluorophenyl)thiourea |
Specifications
458-05-9 | |
Odorless | |
MFCD00041167 | |
UN2811 | |
1-3-fluorophenyl-2-thiourea, 1-3-fluorophenyl thiourea, 3-fluorophenyl thiourea, n-3-fluorophenyl thiourea, amino 3-fluorophenyl amino methane-1-thione, acmc-20apbn, 1-m-fluorophenyl thiourea, 3-fluoro-phenyl-thiourea | |
C1=CC(=CC(=C1)F)NC(=S)N | |
170.205 | |
170.21 | |
N-(3-Fluorophenyl)thiourea |
114°C to 117°C | |
C7H7FN2S | |
1 g | |
2718454 | |
YKOBLHQALWQKTC-UHFFFAOYSA-N | |
(3-fluorophenyl)thiourea | |
2734799 | |
99% |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures