missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(9H-Fluoren-2-ylmethoxycarbonyloxy)succinimide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 243100050

 View more versions of this product

Catalog No. AC243100050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-(9H-Fluoren-2-ylmethoxycarbonyloxy)succinimide
97.5
145.0°C to 153.0°C
0.25% max. (60°C, 2 hrs, 1mm Hg)
97.5% min. (HPLC)
C19H15NO5
5 g
Solubility in water: insoluble
C1CC(=O)N(C1=O)OC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
337.29
337.29
Microfine Powder
82911-69-1
100.0
White
Authentic
Glass bottle
MFCD00010733
fmoc-osu, n-9-fluorenylmethoxycarbonyloxy succinimide, fmoc-onsu, 9-fluorenylmethyl succinimidyl carbonate, fmoc n-hydroxysuccinimide ester, 9h-fluoren-9-yl methyl 2,5-dioxopyrrolidin-1-yl carbonate, 9-fluorenylmethyl n-succinimidyl carbonate, 9-fluorenylmethylsuccinimidyl carbonate, 9-fluorenylmethoxycarbonyl-n-hydroxysuccinimide
WMSUFWLPZLCIHP-UHFFFAOYSA-N
(2,5-dioxopyrrolidin-1-yl) 9H-fluoren-9-ylmethyl carbonate
134122
98%

Chemical Identifiers

82911-69-1
337.29
WMSUFWLPZLCIHP-UHFFFAOYSA-N
134122
C1CC(=O)N(C1=O)OC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
C19H15NO5
MFCD00010733
fmoc-osu, n-9-fluorenylmethoxycarbonyloxy succinimide, fmoc-onsu, 9-fluorenylmethyl succinimidyl carbonate, fmoc n-hydroxysuccinimide ester, 9h-fluoren-9-yl methyl 2,5-dioxopyrrolidin-1-yl carbonate, 9-fluorenylmethyl n-succinimidyl carbonate, 9-fluorenylmethylsuccinimidyl carbonate, 9-fluorenylmethoxycarbonyl-n-hydroxysuccinimide
(2,5-dioxopyrrolidin-1-yl) 9H-fluoren-9-ylmethyl carbonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear eye protection/face protection.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only