Learn More
N-(9H-Fluoren-2-ylmethoxycarbonyloxy)succinimide, 98%, Thermo Scientific Chemicals
$143.44 - $384.82
Chemical Identifiers
CAS | 82911-69-1 |
---|---|
Molecular Formula | C19H15NO5 |
Molecular Weight (g/mol) | 337.29 |
MDL Number | MFCD00010733 |
InChI Key | WMSUFWLPZLCIHP-UHFFFAOYSA-N |
Synonym | fmoc-osu, n-9-fluorenylmethoxycarbonyloxy succinimide, fmoc-onsu, 9-fluorenylmethyl succinimidyl carbonate, fmoc n-hydroxysuccinimide ester, 9h-fluoren-9-yl methyl 2,5-dioxopyrrolidin-1-yl carbonate, 9-fluorenylmethyl n-succinimidyl carbonate, 9-fluorenylmethylsuccinimidyl carbonate, 9-fluorenylmethoxycarbonyl-n-hydroxysuccinimide |
PubChem CID | 134122 |
IUPAC Name | (2,5-dioxopyrrolidin-1-yl) 9H-fluoren-9-ylmethyl carbonate |
SMILES | C1CC(=O)N(C1=O)OC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC243100050
|
Thermo Scientific Chemicals
243100050 |
5 g | Glass bottle |
Each for $143.44
|
|
|||||
AC243100250
|
Thermo Scientific Chemicals
243100250 |
25 g | Glass bottle |
Each for $384.82
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
82911-69-1 | |
337.29 | |
WMSUFWLPZLCIHP-UHFFFAOYSA-N | |
134122 | |
C1CC(=O)N(C1=O)OC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 |
C19H15NO5 | |
MFCD00010733 | |
fmoc-osu, n-9-fluorenylmethoxycarbonyloxy succinimide, fmoc-onsu, 9-fluorenylmethyl succinimidyl carbonate, fmoc n-hydroxysuccinimide ester, 9h-fluoren-9-yl methyl 2,5-dioxopyrrolidin-1-yl carbonate, 9-fluorenylmethyl n-succinimidyl carbonate, 9-fluorenylmethylsuccinimidyl carbonate, 9-fluorenylmethoxycarbonyl-n-hydroxysuccinimide | |
(2,5-dioxopyrrolidin-1-yl) 9H-fluoren-9-ylmethyl carbonate |
Specifications
82911-69-1 | |
100.0 | |
White | |
Authentic | |
Glass bottle | |
MFCD00010733 | |
fmoc-osu, n-9-fluorenylmethoxycarbonyloxy succinimide, fmoc-onsu, 9-fluorenylmethyl succinimidyl carbonate, fmoc n-hydroxysuccinimide ester, 9h-fluoren-9-yl methyl 2,5-dioxopyrrolidin-1-yl carbonate, 9-fluorenylmethyl n-succinimidyl carbonate, 9-fluorenylmethylsuccinimidyl carbonate, 9-fluorenylmethoxycarbonyl-n-hydroxysuccinimide | |
WMSUFWLPZLCIHP-UHFFFAOYSA-N | |
(2,5-dioxopyrrolidin-1-yl) 9H-fluoren-9-ylmethyl carbonate | |
134122 | |
98% | |
N-(9H-Fluoren-2-ylmethoxycarbonyloxy)succinimide |
97.5 | |
145.0°C to 153.0°C | |
0.25% max. (60°C, 2 hrs, 1mm Hg) | |
97.5% min. (HPLC) | |
C19H15NO5 | |
5 g | |
Solubility in water: insoluble | |
C1CC(=O)N(C1=O)OC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 | |
337.29 | |
337.29 | |
Microfine Powder |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Wear eye protection/face protection.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
RUO – Research Use Only