missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-(9H-Fluoren-2-ylmethoxycarbonyloxy)succinimide, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 243100250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N-(9H-Fluoren-2-ylmethoxycarbonyloxy)succinimide | |
97.5 | |
145.0°C to 153.0°C | |
0.25% max. (60°C, 2 hrs, 1mm Hg) | |
97.5% min. (HPLC) | |
C19H15NO5 | |
25 g | |
Solubility in water: insoluble | |
C1CC(=O)N(C1=O)OC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 | |
337.29 | |
337.29 | |
Microfine Powder |
82911-69-1 | |
100.0 | |
White | |
Authentic | |
Glass bottle | |
MFCD00010733 | |
fmoc-osu, n-9-fluorenylmethoxycarbonyloxy succinimide, fmoc-onsu, 9-fluorenylmethyl succinimidyl carbonate, fmoc n-hydroxysuccinimide ester, 9h-fluoren-9-yl methyl 2,5-dioxopyrrolidin-1-yl carbonate, 9-fluorenylmethyl n-succinimidyl carbonate, 9-fluorenylmethylsuccinimidyl carbonate, 9-fluorenylmethoxycarbonyl-n-hydroxysuccinimide | |
WMSUFWLPZLCIHP-UHFFFAOYSA-N | |
(2,5-dioxopyrrolidin-1-yl) 9H-fluoren-9-ylmethyl carbonate | |
134122 | |
98% |
Chemical Identifiers
82911-69-1 | |
337.29 | |
WMSUFWLPZLCIHP-UHFFFAOYSA-N | |
134122 | |
C1CC(=O)N(C1=O)OC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 |
C19H15NO5 | |
MFCD00010733 | |
fmoc-osu, n-9-fluorenylmethoxycarbonyloxy succinimide, fmoc-onsu, 9-fluorenylmethyl succinimidyl carbonate, fmoc n-hydroxysuccinimide ester, 9h-fluoren-9-yl methyl 2,5-dioxopyrrolidin-1-yl carbonate, 9-fluorenylmethyl n-succinimidyl carbonate, 9-fluorenylmethylsuccinimidyl carbonate, 9-fluorenylmethoxycarbonyl-n-hydroxysuccinimide | |
(2,5-dioxopyrrolidin-1-yl) 9H-fluoren-9-ylmethyl carbonate |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Wear eye protection/face protection.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
RUO – Research Use Only