Learn More
N-Acetyl-D-alanine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6345006
Description
N-Acetyl-D-alanine may be used with other D-aminoacylated amino acids as a substrate for the identification, differentiation and characterization of D-aminoacylase(s)/amidohydrolase(s). N-acetyl-D-alanine may be used to study aglycon pocket specific binding on vancomycin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Acetyl-D-alanine may be used with other D-aminoacylated amino acids as a substrate for the identification, differentiation and characterization of D-aminoacylase(s)/amidohydrolase(s). N-acetyl-D-alanine may be used to study aglycon pocket specific binding on vancomycin.
Solubility
Slightly soluble in water.
Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications
N-Acetyl-D-alanine | |
C5H9NO3 | |
n-acetyl-d-alanine, ac-d-ala-oh, r-2-acetamidopropanoic acid, acetyl-d-alanine, n-acetylalanine, 2r-2-acetamidopropanoic acid, r-2-acetylamino-propionic acid, alanine, n-acetyl, n-acetyl-d-ala, n-acetyl d-alanine | |
KTHDTJVBEPMMGL-UHFFFAOYNA-N | |
(2R)-2-acetamidopropanoic acid | |
131.13 | |
131.13 | |
Solid |
19436-52-3 | |
MFCD00063133 | |
Slightly soluble in water. | |
CC(NC(C)=O)C(O)=O | |
5 g | |
132213 | |
98% |
Chemical Identifiers
19436-52-3 | |
131.13 | |
KTHDTJVBEPMMGL-UHFFFAOYNA-N | |
132213 | |
CC(NC(C)=O)C(O)=O |
C5H9NO3 | |
MFCD00063133 | |
n-acetyl-d-alanine, ac-d-ala-oh, r-2-acetamidopropanoic acid, acetyl-d-alanine, n-acetylalanine, 2r-2-acetamidopropanoic acid, r-2-acetylamino-propionic acid, alanine, n-acetyl, n-acetyl-d-ala, n-acetyl d-alanine | |
(2R)-2-acetamidopropanoic acid |
Safety and Handling
EINECSNumber : 243-066-8
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only