missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Acetyl-D-galactosamine, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 434920010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
N-Acetyl-D-galactosamine | |
97.5 | |
Authentic | |
Glass bottle | |
MFCD00136044 | |
85.3 | |
(10% in water) Clear colorless | |
CC(=O)N[C@H]1C(O)O[C@H](CO)[C@H](O)[C@@H]1O | |
221.21 | |
221.21 |
1811-31-0 | |
100.0 | |
97.5% min. (HPLC) | |
C8H15NO6 | |
1g | |
dsstox_cid_28637, dsstox_rid_82907, dsstox_gsid_48711, n-acetyl-, a-d-galactosamine | |
OVRNDRQMDRJTHS-KEWYIRBNSA-N | |
N-[(2R,3S,4S,5S)-3,4,5,6-tetrahydroxy-1-oxohexan-2-yl]acetamide | |
60196345 | |
98% |
Chemical Identifiers
1811-31-0 | |
221.21 | |
OVRNDRQMDRJTHS-KEWYIRBNSA-N | |
60196345 | |
CC(=O)N[C@H]1C(O)O[C@H](CO)[C@H](O)[C@@H]1O |
C8H15NO6 | |
MFCD00136044 | |
dsstox_cid_28637, dsstox_rid_82907, dsstox_gsid_48711, n-acetyl-, a-d-galactosamine | |
N-[(2R,3S,4S,5S)-3,4,5,6-tetrahydroxy-1-oxohexan-2-yl]acetamide |
Safety and Handling
EINECSNumber : 217-321-9
RUO â Research Use Only