missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Acetyl-D-galactosamine, 98%, Thermo Scientific™

Chemical Identifiers

CAS 1811-31-0
Molecular Formula C8H15NO6
Molecular Weight (g/mol) 221.21
MDL Number MFCD00136044
InChI Key OVRNDRQMDRJTHS-KEWYIRBNSA-N
Synonym dsstox_cid_28637, dsstox_rid_82907, dsstox_gsid_48711, n-acetyl-, a-d-galactosamine
PubChem CID 60196345
IUPAC Name N-[(2R,3S,4S,5S)-3,4,5,6-tetrahydroxy-1-oxohexan-2-yl]acetamide
SMILES CC(=O)N[C@H]1C(O)O[C@H](CO)[C@H](O)[C@@H]1O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC434920010
View Documents
Thermo Scientific Chemicals
434920010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC434920050
View Documents
Thermo Scientific Chemicals
434920050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1811-31-0
221.21
OVRNDRQMDRJTHS-KEWYIRBNSA-N
60196345
CC(=O)N[C@H]1C(O)O[C@H](CO)[C@H](O)[C@@H]1O
C8H15NO6
MFCD00136044
dsstox_cid_28637, dsstox_rid_82907, dsstox_gsid_48711, n-acetyl-, a-d-galactosamine
N-[(2R,3S,4S,5S)-3,4,5,6-tetrahydroxy-1-oxohexan-2-yl]acetamide

Specifications

1811-31-0
100.0
97.5% min. (HPLC)
C8H15NO6
1g
dsstox_cid_28637, dsstox_rid_82907, dsstox_gsid_48711, n-acetyl-, a-d-galactosamine
OVRNDRQMDRJTHS-KEWYIRBNSA-N
N-[(2R,3S,4S,5S)-3,4,5,6-tetrahydroxy-1-oxohexan-2-yl]acetamide
60196345
98%
97.5
Authentic
Glass bottle
MFCD00136044
85.3
(10% in water) Clear colorless
CC(=O)N[C@H]1C(O)O[C@H](CO)[C@H](O)[C@@H]1O
221.21
221.21
N-Acetyl-D-galactosamine
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 217-321-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only