missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Acetyl-L-tyrosine ethyl ester monohydrate, 99%, Thermo Scientific Chemicals

$59.41 - $284.40

Chemical Identifiers

CAS 36546-50-6
Molecular Formula C13H19NO5
Molecular Weight (g/mol) 269.30
MDL Number MFCD00149093
InChI Key YWAVLHZJMWEYTA-HVPILOLGNA-N
Synonym n-acetyl-l-tyrosine ethyl ester monohydrate, ethyl n-acetyl-l-tyrosinate hydrate, ac-tyr-oet.h2o, atee, ethyl 2s-2-acetamido-3-4-hydroxyphenyl propanoate hydrate, n-acetyl-l-tyrosineethylestermonohydrate, n-acetyltyrosine ethyl ester monohydrate, ksc491m4n, n-acetyl tyrosine ethyl ester mono hydrate, acetyl-l-tyrosine ethyl ester hydrate
PubChem CID 2723594
IUPAC Name ethyl (2S)-2-acetamido-3-(4-hydroxyphenyl)propanoate;hydrate
SMILES O.CCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(C)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC102750010
SDS View Documents
Thermo Scientific Chemicals
102750010
1 g Glass bottle
Each for $59.41
 
AC102750100
SDS View Documents
Thermo Scientific Chemicals
102750100
10 g Glass bottle
Each for $284.40
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

36546-50-6
269.30
YWAVLHZJMWEYTA-HVPILOLGNA-N
2723594
O.CCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(C)=O
C13H19NO5
MFCD00149093
n-acetyl-l-tyrosine ethyl ester monohydrate, ethyl n-acetyl-l-tyrosinate hydrate, ac-tyr-oet.h2o, atee, ethyl 2s-2-acetamido-3-4-hydroxyphenyl propanoate hydrate, n-acetyl-l-tyrosineethylestermonohydrate, n-acetyltyrosine ethyl ester monohydrate, ksc491m4n, n-acetyl tyrosine ethyl ester mono hydrate, acetyl-l-tyrosine ethyl ester hydrate
ethyl (2S)-2-acetamido-3-(4-hydroxyphenyl)propanoate;hydrate

Specifications

36546-50-6
100.0
White
98.5% min. (HPLC)
C13H19NO5
MFCD00149093
+ 23
Solubility in water: soluble in cold water. Other solubilities: soluble in hot water,soluble in ethanol
O.CCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(C)=O
269.30
269.29
Crystalline Powder
98.5
80°C to 81°C
Authentic
Glass bottle
4-(HO)C6H4CH2CH(NHCOCH3)CO2C2H5·H2O
1 g
n-acetyl-l-tyrosine ethyl ester monohydrate, ethyl n-acetyl-l-tyrosinate hydrate, ac-tyr-oet.h2o, atee, ethyl 2s-2-acetamido-3-4-hydroxyphenyl propanoate hydrate, n-acetyl-l-tyrosineethylestermonohydrate, n-acetyltyrosine ethyl ester monohydrate, ksc491m4n, n-acetyl tyrosine ethyl ester mono hydrate, acetyl-l-tyrosine ethyl ester hydrate
YWAVLHZJMWEYTA-HVPILOLGNA-N
ethyl (2S)-2-acetamido-3-(4-hydroxyphenyl)propanoate;hydrate
2723594
99%
N-Acetyl-L-tyrosine ethyl ester monohydrate
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only