missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Allylphthalimide, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H63724MD
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
N-Allylphthalimide | |
White | |
MFCD00158662 | |
2-allylisoindoline-1,3-dione, n-allylphthalimide, n-2-propenyl phthalimide, 2-allyl-1h-isoindole-1,3 2h-dione, 2-prop-2-en-1-yl-1h-isoindole-1,3 2h-dione, 1h-isoindole-1,3 2h-dione, 2-2-propen-1-yl, 2-prop-2-en-1-yl-2,3-dihydro-1h-isoindole-1,3-dione, n-allylphthalimid, n-allyl-phthalimide, 2-allylisoindole-1,3-dione | |
C=CCN1C(=O)C2=CC=CC=C2C1=O | |
187.20 | |
187.19 | |
Solid |
5428-09-1 | |
C11H9NO2 | |
250 mg | |
MHHGQWMCVNQHLG-UHFFFAOYSA-N | |
2-prop-2-enylisoindole-1,3-dione | |
224401 | |
97% |
Chemical Identifiers
5428-09-1 | |
MFCD00158662 |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures