Learn More
N-Benzoyl-2'-deoxyadenosine, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5230303
Description
Reactant in the synthesis of 2', 5'-Dideoxycytidines and Other Derivatives of 2'-Deoxycytidine. Reactant in the synthesis of Mono- and Diamino Analogues of 2?-Deoxyadenosine, Cordycepin, 9-(3-Deoxy-α-D-Threo-Pentofuranosyl)-Adenine and 9-(2-Deoxy-α-D-Threo-Pentofuranosyl)Adenine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsReactant in the synthesis of 2′, 5′-Dideoxycytidines and Other Derivatives of 2′-Deoxycytidine. Reactant in the synthesis of Mono- and Diamino Analogues of 2′-Deoxyadenosine, Cordycepin, 9-(3-Deoxy-α-D-Threo-Pentofuranosyl)-Adenine and 9-(2-Deoxy-α-D-Threo-Pentofuranosyl)Adenine.
Solubility
Soluble in water (slightly), and methanol (20 mg/ml).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
N-Benzoyl-2'-deoxyadenosine | |
C17H17N5O4 | |
1 g | |
Soluble in water (slightly),and methanol (20mg/ml). | |
OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12 | |
355.35 | |
355.35 |
4546-72-9 | |
MFCD00009628 | |
n6-benzoyl-2'-deoxyadenosine, n-benzoyl-2'-deoxyadenosine, n-benzoyl-2'-deoxy-adenosine, n-9-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purin-6-yl benzamide, n 6-benzoyl-2'-deoxyadenosine, n6-benzoyl-2-deoxyadenosine, adenosine, n-benzoyl-2'-deoxy, n-benzoyldeoxyadenosine, n6-benzoyldeoxyadenosine, 6-n-benzoyldeoxyadenosine | |
PIXHJAPVPCVZSV-LJEKBPCBNA-N | |
N-[9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]purin-6-yl]benzamide | |
107558 | |
≥98% |
Chemical Identifiers
4546-72-9 | |
355.35 | |
PIXHJAPVPCVZSV-LJEKBPCBNA-N | |
N-[9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]purin-6-yl]benzamide |
C17H17N5O4 | |
MFCD00009628 | |
107558 | |
OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12 |
Safety and Handling
EINECSNumber : 224-903-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only