missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Benzoyl-2'-deoxyadenosine, 98+%, Thermo Scientific Chemicals

$193.84 - $759.03

Chemical Identifiers

CAS 4546-72-9
Molecular Formula C17H17N5O4
Molecular Weight (g/mol) 355.35
MDL Number MFCD00009628
InChI Key PIXHJAPVPCVZSV-LJEKBPCBNA-N
PubChem CID 107558
IUPAC Name N-[9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]purin-6-yl]benzamide
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5230303
View Documents
Thermo Scientific Chemicals
H5230303
1 g
Each for $193.84
 
AAH5230306
View Documents
Thermo Scientific Chemicals
H5230306
5 g
Each for $759.03
 
Description

Description

Reactant in the synthesis of 2', 5'-Dideoxycytidines and Other Derivatives of 2'-Deoxycytidine. Reactant in the synthesis of Mono- and Diamino Analogues of 2?-Deoxyadenosine, Cordycepin, 9-(3-Deoxy-α-D-Threo-Pentofuranosyl)-Adenine and 9-(2-Deoxy-α-D-Threo-Pentofuranosyl)Adenine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant in the synthesis of 2′, 5′-Dideoxycytidines and Other Derivatives of 2′-Deoxycytidine. Reactant in the synthesis of Mono- and Diamino Analogues of 2′-Deoxyadenosine, Cordycepin, 9-(3-Deoxy-α-D-Threo-Pentofuranosyl)-Adenine and 9-(2-Deoxy-α-D-Threo-Pentofuranosyl)Adenine.

Solubility
Soluble in water (slightly), and methanol (20 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

4546-72-9
355.35
PIXHJAPVPCVZSV-LJEKBPCBNA-N
N-[9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]purin-6-yl]benzamide
C17H17N5O4
MFCD00009628
107558
OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12

Specifications

4546-72-9
MFCD00009628
n6-benzoyl-2'-deoxyadenosine, n-benzoyl-2'-deoxyadenosine, n-benzoyl-2'-deoxy-adenosine, n-9-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purin-6-yl benzamide, n 6-benzoyl-2'-deoxyadenosine, n6-benzoyl-2-deoxyadenosine, adenosine, n-benzoyl-2'-deoxy, n-benzoyldeoxyadenosine, n6-benzoyldeoxyadenosine, 6-n-benzoyldeoxyadenosine
PIXHJAPVPCVZSV-LJEKBPCBNA-N
N-[9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]purin-6-yl]benzamide
107558
≥98%
C17H17N5O4
1 g
Soluble in water (slightly),and methanol (20mg/ml).
OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12
355.35
355.35
N-Benzoyl-2'-deoxyadenosine
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 224-903-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only