missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Benzoyl-2'-deoxyadenosine, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5230306

 View more versions of this product

Catalog No. AAH5230306


Explore available promotions

Description

Description

Reactant in the synthesis of 2', 5'-Dideoxycytidines and Other Derivatives of 2'-Deoxycytidine. Reactant in the synthesis of Mono- and Diamino Analogues of 2?-Deoxyadenosine, Cordycepin, 9-(3-Deoxy-α-D-Threo-Pentofuranosyl)-Adenine and 9-(2-Deoxy-α-D-Threo-Pentofuranosyl)Adenine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant in the synthesis of 2′, 5′-Dideoxycytidines and Other Derivatives of 2′-Deoxycytidine. Reactant in the synthesis of Mono- and Diamino Analogues of 2′-Deoxyadenosine, Cordycepin, 9-(3-Deoxy-α-D-Threo-Pentofuranosyl)-Adenine and 9-(2-Deoxy-α-D-Threo-Pentofuranosyl)Adenine.

Solubility
Soluble in water (slightly), and methanol (20 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

N-Benzoyl-2'-deoxyadenosine
C17H17N5O4
5 g
Soluble in water (slightly),and methanol (20mg/ml).
OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12
355.35
355.35
4546-72-9
MFCD00009628
n6-benzoyl-2'-deoxyadenosine, n-benzoyl-2'-deoxyadenosine, n-benzoyl-2'-deoxy-adenosine, n-9-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purin-6-yl benzamide, n 6-benzoyl-2'-deoxyadenosine, n6-benzoyl-2-deoxyadenosine, adenosine, n-benzoyl-2'-deoxy, n-benzoyldeoxyadenosine, n6-benzoyldeoxyadenosine, 6-n-benzoyldeoxyadenosine
PIXHJAPVPCVZSV-LJEKBPCBNA-N
N-[9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]purin-6-yl]benzamide
107558
≥98%

Chemical Identifiers

4546-72-9
355.35
PIXHJAPVPCVZSV-LJEKBPCBNA-N
N-[9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]purin-6-yl]benzamide
C17H17N5O4
MFCD00009628
107558
OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 224-903-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only