missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Benzyl-p-toluenesulfonamide, Thermo Scientific Chemicals

A potent inhibitor of calcium-stimulated myosin S1 ATPase

Supplier:  Thermo Scientific Chemicals J64910MA

 View more versions of this product

Catalog No. AAJ64910MA


Explore available promotions

Description

Description

A potent inhibitor of calcium-stimulated myosin S1 ATPase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-Benzyl-p-toluenesulfonamide
115°C
C14H15NO2S
10 mg
n-benzyl-p-toluenesulfonamide, n-tosylbenzylamine, p-toluenesulfonamide, n-benzyl, n-benzyl-p-toluenesulphonamide, n-benzyl-4-methyl-benzenesulfonamide, n-benzyl-4-methylbenzene-1-sulfonamide, n-benzyl-p-toluene sulphonamide, benzenesulfonamide, 4-methyl-n-phenylmethyl, 4-methyl-n-phenylmethyl benzenesulfonamide, 4-methyl-n-phenylmethyl-benzenesulfonamid
CC1=CC=C(C=C1)S(=O)(=O)NCC2=CC=CC=C2
261.339
261.34
1576-37-0
White
MFCD00159328
Light sensitive
WTHKAJZQYNKTCJ-UHFFFAOYSA-N
N-benzyl-4-methylbenzenesulfonamide
95801
Crystalline Powder

Chemical Identifiers

1576-37-0
261.339
WTHKAJZQYNKTCJ-UHFFFAOYSA-N
95801
CC1=CC=C(C=C1)S(=O)(=O)NCC2=CC=CC=C2
C14H15NO2S
MFCD00159328
n-benzyl-p-toluenesulfonamide, n-tosylbenzylamine, p-toluenesulfonamide, n-benzyl, n-benzyl-p-toluenesulphonamide, n-benzyl-4-methyl-benzenesulfonamide, n-benzyl-4-methylbenzene-1-sulfonamide, n-benzyl-p-toluene sulphonamide, benzenesulfonamide, 4-methyl-n-phenylmethyl, 4-methyl-n-phenylmethyl benzenesulfonamide, 4-methyl-n-phenylmethyl-benzenesulfonamid
N-benzyl-4-methylbenzenesulfonamide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

RTECSNumber : XT5490000

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only