Learn More
N-Benzyl-p-toluenesulfonamide, Thermo Scientific Chemicals
A potent inhibitor of calcium-stimulated myosin S1 ATPase
$231.75 - $366.55
Chemical Identifiers
CAS | 1576-37-0 |
---|---|
Molecular Formula | C14H15NO2S |
Molecular Weight (g/mol) | 261.339 |
MDL Number | MFCD00159328 |
InChI Key | WTHKAJZQYNKTCJ-UHFFFAOYSA-N |
Synonym | n-benzyl-p-toluenesulfonamide, n-tosylbenzylamine, p-toluenesulfonamide, n-benzyl, n-benzyl-p-toluenesulphonamide, n-benzyl-4-methyl-benzenesulfonamide, n-benzyl-4-methylbenzene-1-sulfonamide, n-benzyl-p-toluene sulphonamide, benzenesulfonamide, 4-methyl-n-phenylmethyl, 4-methyl-n-phenylmethyl benzenesulfonamide, 4-methyl-n-phenylmethyl-benzenesulfonamid |
PubChem CID | 95801 |
IUPAC Name | N-benzyl-4-methylbenzenesulfonamide |
SMILES | CC1=CC=C(C=C1)S(=O)(=O)NCC2=CC=CC=C2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ64910LB0
|
Thermo Scientific Chemicals
J64910LB0 |
5 mg |
Each for $231.75
|
|
|||||
AAJ64910MA
|
Thermo Scientific Chemicals
J64910MA |
10 mg |
Each for $366.55
|
|
|||||
Description
A potent inhibitor of calcium-stimulated myosin S1 ATPase
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1576-37-0 | |
261.339 | |
WTHKAJZQYNKTCJ-UHFFFAOYSA-N | |
95801 | |
CC1=CC=C(C=C1)S(=O)(=O)NCC2=CC=CC=C2 |
C14H15NO2S | |
MFCD00159328 | |
n-benzyl-p-toluenesulfonamide, n-tosylbenzylamine, p-toluenesulfonamide, n-benzyl, n-benzyl-p-toluenesulphonamide, n-benzyl-4-methyl-benzenesulfonamide, n-benzyl-4-methylbenzene-1-sulfonamide, n-benzyl-p-toluene sulphonamide, benzenesulfonamide, 4-methyl-n-phenylmethyl, 4-methyl-n-phenylmethyl benzenesulfonamide, 4-methyl-n-phenylmethyl-benzenesulfonamid | |
N-benzyl-4-methylbenzenesulfonamide |
Specifications
1576-37-0 | |
White | |
MFCD00159328 | |
Light sensitive | |
WTHKAJZQYNKTCJ-UHFFFAOYSA-N | |
N-benzyl-4-methylbenzenesulfonamide | |
95801 | |
Crystalline Powder |
115°C | |
C14H15NO2S | |
5 mg | |
n-benzyl-p-toluenesulfonamide, n-tosylbenzylamine, p-toluenesulfonamide, n-benzyl, n-benzyl-p-toluenesulphonamide, n-benzyl-4-methyl-benzenesulfonamide, n-benzyl-4-methylbenzene-1-sulfonamide, n-benzyl-p-toluene sulphonamide, benzenesulfonamide, 4-methyl-n-phenylmethyl, 4-methyl-n-phenylmethyl benzenesulfonamide, 4-methyl-n-phenylmethyl-benzenesulfonamid | |
CC1=CC=C(C=C1)S(=O)(=O)NCC2=CC=CC=C2 | |
261.339 | |
261.34 | |
N-Benzyl-p-toluenesulfonamide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
RTECSNumber : XT5490000
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only