missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Boc-2-cyclopropyl-L-glycine, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H62914MD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N-Boc-2-cyclopropyl-L-glycine | |
White | |
MFCD06659132,MFCD01861823 | |
boc-l-cyclopropylglycine, s-2-tert-butoxycarbonyl amino-2-cyclopropylacetic acid, boc-s-cyclopropylglycine, s-tert-butoxycarbonylamino-cyclopropyl-acetic acid, s-boc-amino-cyclopropylacetic acid, 2s-2-amino-2-cyclopropylethanoic acid, n-boc protected, s-2-tert-butoxycarbonylamino-2-cyclopropylacetic acid, s-tert-butoxycarbonyl amino cyclopropyl acetic acid, boc-l-cyclopropyl glycine | |
CC(C)(C)OC(=O)N[C@@H](C1CC1)C(O)=O | |
250 mg | |
2761461 | |
95% |
155976-13-9 | |
C10H17NO4 | |
6650020 | |
QFVJNEASAAJIDF-ZETCQYMHSA-N | |
(2S)-2-cyclopropyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid | |
215.25 | |
215.25 | |
Powder |
Chemical Identifiers
155976-13-9 | |
215.25 | |
QFVJNEASAAJIDF-ZETCQYMHSA-N | |
2761461 | |
CC(C)(C)OC(=O)N[C@@H](C1CC1)C(O)=O |
C10H17NO4 | |
MFCD06659132,MFCD01861823 | |
boc-l-cyclopropylglycine, s-2-tert-butoxycarbonyl amino-2-cyclopropylacetic acid, boc-s-cyclopropylglycine, s-tert-butoxycarbonylamino-cyclopropyl-acetic acid, s-boc-amino-cyclopropylacetic acid, 2s-2-amino-2-cyclopropylethanoic acid, n-boc protected, s-2-tert-butoxycarbonylamino-2-cyclopropylacetic acid, s-tert-butoxycarbonyl amino cyclopropyl acetic acid, boc-l-cyclopropyl glycine | |
(2S)-2-cyclopropyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only