missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Boc-2-cyclopropyl-L-glycine, 95%, Thermo Scientific Chemicals

$123.26 - $261.14

Chemical Identifiers

CAS 155976-13-9
Molecular Formula C10H17NO4
Molecular Weight (g/mol) 215.25
MDL Number MFCD06659132,MFCD01861823
InChI Key QFVJNEASAAJIDF-ZETCQYMHSA-N
Synonym boc-l-cyclopropylglycine, s-2-tert-butoxycarbonyl amino-2-cyclopropylacetic acid, boc-s-cyclopropylglycine, s-tert-butoxycarbonylamino-cyclopropyl-acetic acid, s-boc-amino-cyclopropylacetic acid, 2s-2-amino-2-cyclopropylethanoic acid, n-boc protected, s-2-tert-butoxycarbonylamino-2-cyclopropylacetic acid, s-tert-butoxycarbonyl amino cyclopropyl acetic acid, boc-l-cyclopropyl glycine
PubChem CID 2761461
IUPAC Name (2S)-2-cyclopropyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid
SMILES CC(C)(C)OC(=O)N[C@@H](C1CC1)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62914MD
View Documents
Thermo Scientific Chemicals
H62914MD
250 mg
Each for $123.26
 
AAH6291403
View Documents
Thermo Scientific Chemicals
H6291403
1 g
Each for $261.14
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

155976-13-9
215.25
QFVJNEASAAJIDF-ZETCQYMHSA-N
2761461
CC(C)(C)OC(=O)N[C@@H](C1CC1)C(O)=O
C10H17NO4
MFCD06659132,MFCD01861823
boc-l-cyclopropylglycine, s-2-tert-butoxycarbonyl amino-2-cyclopropylacetic acid, boc-s-cyclopropylglycine, s-tert-butoxycarbonylamino-cyclopropyl-acetic acid, s-boc-amino-cyclopropylacetic acid, 2s-2-amino-2-cyclopropylethanoic acid, n-boc protected, s-2-tert-butoxycarbonylamino-2-cyclopropylacetic acid, s-tert-butoxycarbonyl amino cyclopropyl acetic acid, boc-l-cyclopropyl glycine
(2S)-2-cyclopropyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid

Specifications

155976-13-9
C10H17NO4
6650020
QFVJNEASAAJIDF-ZETCQYMHSA-N
(2S)-2-cyclopropyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid
215.25
215.25
Powder
White
MFCD06659132,MFCD01861823
boc-l-cyclopropylglycine, s-2-tert-butoxycarbonyl amino-2-cyclopropylacetic acid, boc-s-cyclopropylglycine, s-tert-butoxycarbonylamino-cyclopropyl-acetic acid, s-boc-amino-cyclopropylacetic acid, 2s-2-amino-2-cyclopropylethanoic acid, n-boc protected, s-2-tert-butoxycarbonylamino-2-cyclopropylacetic acid, s-tert-butoxycarbonyl amino cyclopropyl acetic acid, boc-l-cyclopropyl glycine
CC(C)(C)OC(=O)N[C@@H](C1CC1)C(O)=O
250 mg
2761461
95%
N-Boc-2-cyclopropyl-L-glycine
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only