missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-BOC-Glycine methyleester, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 399380250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
N-BOC-Glycine methyl ester | |
96.0 | |
97% | |
MFCD00038267 | |
boc-glycine methyl ester, boc-gly-ome, n-tert-butoxycarbonyl glycine methyl ester, n-boc-glycine methyl ester, boc-glycinemethylester, methyl 2-tert-butoxycarbonyl amino acetate, methyl n-tert-butoxycarbonyl glycinate, glycine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl tert-butoxycarbonyl amino acetate | |
CC(C)(C)OC(=O)NCC(=O)OC | |
189.211 | |
189.21 | |
1.2800g/mL | |
109°C |
31954-27-5 | |
100.0 | |
C8H15NO4 | |
Glass bottle | |
PHUZOEOLWIHIKH-UHFFFAOYSA-N | |
methyl 2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate | |
637609 | |
97% | |
25mL | |
1.28 |
Chemical Identifiers
31954-27-5 | |
189.211 | |
PHUZOEOLWIHIKH-UHFFFAOYSA-N | |
637609 | |
CC(C)(C)OC(=O)NCC(=O)OC |
C8H15NO4 | |
MFCD00038267 | |
boc-glycine methyl ester, boc-gly-ome, n-tert-butoxycarbonyl glycine methyl ester, n-boc-glycine methyl ester, boc-glycinemethylester, methyl 2-tert-butoxycarbonyl amino acetate, methyl n-tert-butoxycarbonyl glycinate, glycine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl tert-butoxycarbonyl amino acetate | |
methyl 2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate |
RUO â Research Use Only