missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-BOC-Glycine methyleester, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 31954-27-5 |
---|---|
Molecular Formula | C8H15NO4 |
Molecular Weight (g/mol) | 189.211 |
MDL Number | MFCD00038267 |
InChI Key | PHUZOEOLWIHIKH-UHFFFAOYSA-N |
Synonym | boc-glycine methyl ester, boc-gly-ome, n-tert-butoxycarbonyl glycine methyl ester, n-boc-glycine methyl ester, boc-glycinemethylester, methyl 2-tert-butoxycarbonyl amino acetate, methyl n-tert-butoxycarbonyl glycinate, glycine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl tert-butoxycarbonyl amino acetate |
PubChem CID | 637609 |
IUPAC Name | methyl 2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate |
SMILES | CC(C)(C)OC(=O)NCC(=O)OC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC399380250
|
Thermo Scientific Chemicals
399380250 |
25mL | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
31954-27-5 | |
189.211 | |
PHUZOEOLWIHIKH-UHFFFAOYSA-N | |
637609 | |
CC(C)(C)OC(=O)NCC(=O)OC |
C8H15NO4 | |
MFCD00038267 | |
boc-glycine methyl ester, boc-gly-ome, n-tert-butoxycarbonyl glycine methyl ester, n-boc-glycine methyl ester, boc-glycinemethylester, methyl 2-tert-butoxycarbonyl amino acetate, methyl n-tert-butoxycarbonyl glycinate, glycine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl tert-butoxycarbonyl amino acetate | |
methyl 2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate |
Specifications
31954-27-5 | |
100.0 | |
C8H15NO4 | |
Glass bottle | |
PHUZOEOLWIHIKH-UHFFFAOYSA-N | |
methyl 2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate | |
637609 | |
97% | |
25mL | |
1.28 |
96.0 | |
97% | |
MFCD00038267 | |
boc-glycine methyl ester, boc-gly-ome, n-tert-butoxycarbonyl glycine methyl ester, n-boc-glycine methyl ester, boc-glycinemethylester, methyl 2-tert-butoxycarbonyl amino acetate, methyl n-tert-butoxycarbonyl glycinate, glycine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl tert-butoxycarbonyl amino acetate | |
CC(C)(C)OC(=O)NCC(=O)OC | |
189.211 | |
189.21 | |
1.2800g/mL | |
109°C | |
N-BOC-Glycine methyl ester |
RUO â Research Use Only