missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Boc-L-isoleucine, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1602114

 View more versions of this product

Catalog No. AAA1602114


Explore available promotions

Description

Description

N-Boc-L-isoleucine is used to prepare BOC-L-isoleucine hydroxysuccinimide ester by using 1-hydroxybenzotriazole and 1-ethyl-3-<3-(dimethylamino)propyl>carbodiimide hydrochloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-isoleucine is used to prepare BOC-L-isoleucine hydroxysuccinimide ester by using 1-hydroxybenzotriazole and 1-ethyl-3-<3-(dimethylamino)propyl>carbodiimide hydrochloride.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong acids and strong bases. Store in a cool place.
Specifications

Specifications

N-Boc-L-isoleucine
67°C to 70°C
MFCD00038324 MFCD00237550
1711700
Soluble in methanol. Insoluble in water.
CC[C@H](C)[C@H](NC(=O)OC(C)(C)C)C(O)=O
231.29
231.06
13139-16-7
C11H21NO4
25 g
boc-l-isoleucine, boc-ile-oh, n-boc-l-isoleucine, n-tert-butoxycarbonyl-l-isoleucine, boc-dl-ile-oh, boc-l-ile-oh, 2s,3s-2-tert-butoxycarbonyl amino-3-methylpentanoic acid, l-isoleucine, n-1,1-dimethylethoxy carbonyl, boc-isoleucine, n-t-boc-l-isoleucine
QJCNLJWUIOIMMF-YUMQZZPRSA-N
(2S,3S)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
2724762
≥98%

Chemical Identifiers

13139-16-7
231.29
QJCNLJWUIOIMMF-YUMQZZPRSA-N
2724762
CC[C@H](C)[C@H](NC(=O)OC(C)(C)C)C(O)=O
C11H21NO4
MFCD00038324 MFCD00237550
boc-l-isoleucine, boc-ile-oh, n-boc-l-isoleucine, n-tert-butoxycarbonyl-l-isoleucine, boc-dl-ile-oh, boc-l-ile-oh, 2s,3s-2-tert-butoxycarbonyl amino-3-methylpentanoic acid, l-isoleucine, n-1,1-dimethylethoxy carbonyl, boc-isoleucine, n-t-boc-l-isoleucine
(2S,3S)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 236-074-8

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only