missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Boc-L-methionine sulfoxide, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6305306
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
N-Boc-L-methionine sulfoxide | |
C10H19NO5S | |
boc-met o-oh, boc-l-methionine sulfoxide, 2s-2-tert-butoxycarbonyl amino-4-methanesulfinylbutanoic acid, boc-met o, pubchem12247, n-alpha-t-butoxycarbonyl-l-methioninesulfoxid, n-alpha-tert-butyloxycarbonyl-l-methioninesulfoxide, 2s-2-tert-butoxycarbonylamino-4-methylsulfinyl-butanoic acid, 2s-2-tert-butoxy carbonyl amino-4-methanesulfinylbutanoic acid, 2s-2-2-methylpropan-2-yl oxycarbonylamino-4-methylsulfinylbutanoic acid | |
CS(=O)CC[C@H](NC(=O)OC(C)(C)C)C(O)=O | |
5 g | |
7408090 | |
98% |
34805-21-5 | |
MFCD00037172 | |
FVSDTYGQCVACMH-ISJKBYAMSA-N | |
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-methylsulfinylbutanoic acid | |
265.32 | |
265.33 | |
Solid |
Chemical Identifiers
34805-21-5 | |
265.32 | |
FVSDTYGQCVACMH-ISJKBYAMSA-N | |
7408090 | |
CS(=O)CC[C@H](NC(=O)OC(C)(C)C)C(O)=O |
C10H19NO5S | |
MFCD00037172 | |
boc-met o-oh, boc-l-methionine sulfoxide, 2s-2-tert-butoxycarbonyl amino-4-methanesulfinylbutanoic acid, boc-met o, pubchem12247, n-alpha-t-butoxycarbonyl-l-methioninesulfoxid, n-alpha-tert-butyloxycarbonyl-l-methioninesulfoxide, 2s-2-tert-butoxycarbonylamino-4-methylsulfinyl-butanoic acid, 2s-2-tert-butoxy carbonyl amino-4-methanesulfinylbutanoic acid, 2s-2-2-methylpropan-2-yl oxycarbonylamino-4-methylsulfinylbutanoic acid | |
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-methylsulfinylbutanoic acid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold