missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Boc-L-methionine sulfoxide, 98%, Thermo Scientific™

$91.52 - $336.04

Chemical Identifiers

CAS 34805-21-5
Molecular Formula C10H19NO5S
Molecular Weight (g/mol) 265.32
MDL Number MFCD00037172
InChI Key FVSDTYGQCVACMH-ISJKBYAMSA-N
Synonym boc-met o-oh, boc-l-methionine sulfoxide, 2s-2-tert-butoxycarbonyl amino-4-methanesulfinylbutanoic acid, boc-met o, pubchem12247, n-alpha-t-butoxycarbonyl-l-methioninesulfoxid, n-alpha-tert-butyloxycarbonyl-l-methioninesulfoxide, 2s-2-tert-butoxycarbonylamino-4-methylsulfinyl-butanoic acid, 2s-2-tert-butoxy carbonyl amino-4-methanesulfinylbutanoic acid, 2s-2-2-methylpropan-2-yl oxycarbonylamino-4-methylsulfinylbutanoic acid
PubChem CID 7408090
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-methylsulfinylbutanoic acid
SMILES CS(=O)CC[C@H](NC(=O)OC(C)(C)C)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6305306
View Documents
Thermo Scientific Chemicals
H6305306
5 g
Each for $91.52
 
AAH6305314
View Documents
Thermo Scientific Chemicals
H6305314
25 g
Each for $336.04
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

34805-21-5
265.32
FVSDTYGQCVACMH-ISJKBYAMSA-N
7408090
CS(=O)CC[C@H](NC(=O)OC(C)(C)C)C(O)=O
C10H19NO5S
MFCD00037172
boc-met o-oh, boc-l-methionine sulfoxide, 2s-2-tert-butoxycarbonyl amino-4-methanesulfinylbutanoic acid, boc-met o, pubchem12247, n-alpha-t-butoxycarbonyl-l-methioninesulfoxid, n-alpha-tert-butyloxycarbonyl-l-methioninesulfoxide, 2s-2-tert-butoxycarbonylamino-4-methylsulfinyl-butanoic acid, 2s-2-tert-butoxy carbonyl amino-4-methanesulfinylbutanoic acid, 2s-2-2-methylpropan-2-yl oxycarbonylamino-4-methylsulfinylbutanoic acid
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-methylsulfinylbutanoic acid

Specifications

34805-21-5
MFCD00037172
FVSDTYGQCVACMH-ISJKBYAMSA-N
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-methylsulfinylbutanoic acid
265.32
265.33
Solid
C10H19NO5S
boc-met o-oh, boc-l-methionine sulfoxide, 2s-2-tert-butoxycarbonyl amino-4-methanesulfinylbutanoic acid, boc-met o, pubchem12247, n-alpha-t-butoxycarbonyl-l-methioninesulfoxid, n-alpha-tert-butyloxycarbonyl-l-methioninesulfoxide, 2s-2-tert-butoxycarbonylamino-4-methylsulfinyl-butanoic acid, 2s-2-tert-butoxy carbonyl amino-4-methanesulfinylbutanoic acid, 2s-2-2-methylpropan-2-yl oxycarbonylamino-4-methylsulfinylbutanoic acid
CS(=O)CC[C@H](NC(=O)OC(C)(C)C)C(O)=O
5 g
7408090
98%
N-Boc-L-methionine sulfoxide
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'