missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Boc-L-methionine sulfoxide, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H6305314

 View more versions of this product

Catalog No. AAH6305314


Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

N-Boc-L-methionine sulfoxide
C10H19NO5S
boc-met o-oh, boc-l-methionine sulfoxide, 2s-2-tert-butoxycarbonyl amino-4-methanesulfinylbutanoic acid, boc-met o, pubchem12247, n-alpha-t-butoxycarbonyl-l-methioninesulfoxid, n-alpha-tert-butyloxycarbonyl-l-methioninesulfoxide, 2s-2-tert-butoxycarbonylamino-4-methylsulfinyl-butanoic acid, 2s-2-tert-butoxy carbonyl amino-4-methanesulfinylbutanoic acid, 2s-2-2-methylpropan-2-yl oxycarbonylamino-4-methylsulfinylbutanoic acid
CS(=O)CC[C@H](NC(=O)OC(C)(C)C)C(O)=O
25 g
7408090
98%
34805-21-5
MFCD00037172
FVSDTYGQCVACMH-ISJKBYAMSA-N
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-methylsulfinylbutanoic acid
265.32
265.33
Solid

Chemical Identifiers

34805-21-5
265.32
FVSDTYGQCVACMH-ISJKBYAMSA-N
7408090
CS(=O)CC[C@H](NC(=O)OC(C)(C)C)C(O)=O
C10H19NO5S
MFCD00037172
boc-met o-oh, boc-l-methionine sulfoxide, 2s-2-tert-butoxycarbonyl amino-4-methanesulfinylbutanoic acid, boc-met o, pubchem12247, n-alpha-t-butoxycarbonyl-l-methioninesulfoxid, n-alpha-tert-butyloxycarbonyl-l-methioninesulfoxide, 2s-2-tert-butoxycarbonylamino-4-methylsulfinyl-butanoic acid, 2s-2-tert-butoxy carbonyl amino-4-methanesulfinylbutanoic acid, 2s-2-2-methylpropan-2-yl oxycarbonylamino-4-methylsulfinylbutanoic acid
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-methylsulfinylbutanoic acid
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions