missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Boc-(methylamino)acetaldehyde, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5087003

 View more versions of this product

Catalog No. AAH5087003


Explore available promotions

Description

Description

N-Boc-(methylamino)acetaldehyde is involved in the preparation of benzo[d]imidazole inhibitor of co-activator associated arginine methyltransferase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-(methylamino)acetaldehyde is involved in the preparation of benzo[d]imidazole inhibitor of co-activator associated arginine methyltransferase.

Solubility
Slightly miscible with water.

Notes
Air sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
Specifications

Specifications

N-Boc-(methylamino)acetaldehyde
C8H15NO3
n-boc-methylamino acetaldehyde, tert-butyl methyl 2-oxoethyl carbamate, tert-butyl n-methyl-n-2-oxoethyl carbamate, n-tert-butoxycarbonyl-n-methylaminoacetaldehyde, methyl-2-oxo-ethyl-carbamic acid tert-butyl ester, carbamic acid, methyl 2-oxoethyl-, 1,1-dimethylethyl ester, carbamic acid,n-methyl-n-2-oxoethyl-, 1,1-dimethylethyl ester, acmc-20a2gd, n-t-butoxycarbonylsarcosinal, n-boc-n-methyl-2-amino-acetaldehyde
MSWTVSDFEYSRMQ-UHFFFAOYSA-N
tert-butyl N-methyl-N-(2-oxoethyl)carbamate
173.212
173.21
98%
123387-72-4
MFCD08064267
Slightly miscible with water.
CC(C)(C)OC(=O)N(C)CC=O
1 g
10725881
Air sensitive

Chemical Identifiers

123387-72-4
173.212
MSWTVSDFEYSRMQ-UHFFFAOYSA-N
10725881
CC(C)(C)OC(=O)N(C)CC=O
C8H15NO3
MFCD08064267
n-boc-methylamino acetaldehyde, tert-butyl methyl 2-oxoethyl carbamate, tert-butyl n-methyl-n-2-oxoethyl carbamate, n-tert-butoxycarbonyl-n-methylaminoacetaldehyde, methyl-2-oxo-ethyl-carbamic acid tert-butyl ester, carbamic acid, methyl 2-oxoethyl-, 1,1-dimethylethyl ester, carbamic acid,n-methyl-n-2-oxoethyl-, 1,1-dimethylethyl ester, acmc-20a2gd, n-t-butoxycarbonylsarcosinal, n-boc-n-methyl-2-amino-acetaldehyde
tert-butyl N-methyl-N-(2-oxoethyl)carbamate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only