Learn More
N-Boc-(methylamino)acetaldehyde, 97%, Thermo Scientific Chemicals
$158.63 - $532.42
Chemical Identifiers
CAS | 123387-72-4 |
---|---|
Molecular Formula | C8H15NO3 |
Molecular Weight (g/mol) | 173.212 |
MDL Number | MFCD08064267 |
InChI Key | MSWTVSDFEYSRMQ-UHFFFAOYSA-N |
Synonym | n-boc-methylamino acetaldehyde, tert-butyl methyl 2-oxoethyl carbamate, tert-butyl n-methyl-n-2-oxoethyl carbamate, n-tert-butoxycarbonyl-n-methylaminoacetaldehyde, methyl-2-oxo-ethyl-carbamic acid tert-butyl ester, carbamic acid, methyl 2-oxoethyl-, 1,1-dimethylethyl ester, carbamic acid,n-methyl-n-2-oxoethyl-, 1,1-dimethylethyl ester, acmc-20a2gd, n-t-butoxycarbonylsarcosinal, n-boc-n-methyl-2-amino-acetaldehyde |
PubChem CID | 10725881 |
IUPAC Name | tert-butyl N-methyl-N-(2-oxoethyl)carbamate |
SMILES | CC(C)(C)OC(=O)N(C)CC=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH50870MD
|
Thermo Scientific Chemicals
H50870MD |
250 mg |
Each for $158.63
|
|
|||||
AAH5087003
|
Thermo Scientific Chemicals
H5087003 |
1 g |
Each for $532.42
|
|
|||||
Description
N-Boc-(methylamino)acetaldehyde is involved in the preparation of benzo[d]imidazole inhibitor of co-activator associated arginine methyltransferase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Boc-(methylamino)acetaldehyde is involved in the preparation of benzo[d]imidazole inhibitor of co-activator associated arginine methyltransferase.
Solubility
Slightly miscible with water.
Notes
Air sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
Chemical Identifiers
123387-72-4 | |
173.212 | |
MSWTVSDFEYSRMQ-UHFFFAOYSA-N | |
10725881 | |
CC(C)(C)OC(=O)N(C)CC=O |
C8H15NO3 | |
MFCD08064267 | |
n-boc-methylamino acetaldehyde, tert-butyl methyl 2-oxoethyl carbamate, tert-butyl n-methyl-n-2-oxoethyl carbamate, n-tert-butoxycarbonyl-n-methylaminoacetaldehyde, methyl-2-oxo-ethyl-carbamic acid tert-butyl ester, carbamic acid, methyl 2-oxoethyl-, 1,1-dimethylethyl ester, carbamic acid,n-methyl-n-2-oxoethyl-, 1,1-dimethylethyl ester, acmc-20a2gd, n-t-butoxycarbonylsarcosinal, n-boc-n-methyl-2-amino-acetaldehyde | |
tert-butyl N-methyl-N-(2-oxoethyl)carbamate |
Specifications
123387-72-4 | |
MFCD08064267 | |
Slightly miscible with water. | |
CC(C)(C)OC(=O)N(C)CC=O | |
173.212 | |
173.21 | |
250 mg | |
N-Boc-(methylamino)acetaldehyde |
C8H15NO3 | |
n-boc-methylamino acetaldehyde, tert-butyl methyl 2-oxoethyl carbamate, tert-butyl n-methyl-n-2-oxoethyl carbamate, n-tert-butoxycarbonyl-n-methylaminoacetaldehyde, methyl-2-oxo-ethyl-carbamic acid tert-butyl ester, carbamic acid, methyl 2-oxoethyl-, 1,1-dimethylethyl ester, carbamic acid,n-methyl-n-2-oxoethyl-, 1,1-dimethylethyl ester, acmc-20a2gd, n-t-butoxycarbonylsarcosinal, n-boc-n-methyl-2-amino-acetaldehyde | |
MSWTVSDFEYSRMQ-UHFFFAOYSA-N | |
tert-butyl N-methyl-N-(2-oxoethyl)carbamate | |
10725881 | |
98% | |
Air sensitive |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only