Learn More
n-Butyl propionate, 99+%, Thermo Scientific Chemicals
$265.72 - $265.72
Chemical Identifiers
CAS | 590-01-2 |
---|---|
Molecular Formula | C7H14O2 |
Molecular Weight (g/mol) | 130.19 |
MDL Number | MFCD00009448 |
InChI Key | BTMVHUNTONAYDX-UHFFFAOYSA-N |
Synonym | butyl propionate, n-butyl propionate, propanoic acid, butyl ester, n-butyl propanoate, propionic acid, butyl ester, propionic acid butyl ester, unii-2nxc4ak99e, butyl propionate natural, fema no. 2211, n-butyl n-propionate |
PubChem CID | 11529 |
IUPAC Name | butyl propanoate |
SMILES | CCCCOC(=O)CC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC269480010
|
Thermo Scientific Chemicals
269480010 |
1 L | Glass bottle |
Each for $265.72
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
590-01-2 | |
130.19 | |
BTMVHUNTONAYDX-UHFFFAOYSA-N | |
11529 | |
CCCCOC(=O)CC |
C7H14O2 | |
MFCD00009448 | |
butyl propionate, n-butyl propionate, propanoic acid, butyl ester, n-butyl propanoate, propionic acid, butyl ester, propionic acid butyl ester, unii-2nxc4ak99e, butyl propionate natural, fema no. 2211, n-butyl n-propionate | |
butyl propanoate |
Specifications
590-01-2 | |
100.0 | |
Colorless | |
145°C | |
Authentic | |
Glass bottle | |
1.4000 to 1.402 | |
MFCD00009448 | |
02, 241 | |
15, 1590 | |
Solubility in water: 2g/L (20°C). Other solubilities: soluble in alcohol and ether | |
CCCCOC(=O)CC | |
130.19 | |
0.83 mPa.s (20°C) | |
99+% | |
n-Butyl propionate |
99.0 | |
-75°C | |
0.8800g/mL | |
38°C | |
99+% | |
C7H14O2 | |
CH3(CH2)3OC(O)C2H5 | |
1 L | |
0.88 | |
butyl propionate, n-butyl propionate, propanoic acid, butyl ester, n-butyl propanoate, propionic acid, butyl ester, propionic acid butyl ester, unii-2nxc4ak99e, butyl propionate natural, fema no. 2211, n-butyl n-propionate | |
BTMVHUNTONAYDX-UHFFFAOYSA-N | |
butyl propanoate | |
11529 | |
130.19 | |
Liquid |
Safety and Handling
GHS H Statement:
Flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 209-669-5
RUO – Research Use Only