missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Butyl propionate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2012536

 View more versions of this product

Catalog No. AAB2012536


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

n-Butyl propionate
0.87
38°C (101°F)
1.402
500 g
14,1587
BTMVHUNTONAYDX-UHFFFAOYSA-N
butyl propanoate
11529
99%
590-01-2
144°C to 146°C
C7H14O2
MFCD00009448
UN3272
butyl propionate, n-butyl propionate, propanoic acid, butyl ester, n-butyl propanoate, propionic acid, butyl ester, propionic acid butyl ester, unii-2nxc4ak99e, butyl propionate natural, fema no. 2211, n-butyl n-propionate
CCCCOC(=O)CC
130.187
130.19

Chemical Identifiers

590-01-2
130.187
BTMVHUNTONAYDX-UHFFFAOYSA-N
11529
CCCCOC(=O)CC
C7H14O2
MFCD00009448
butyl propionate, n-butyl propionate, propanoic acid, butyl ester, n-butyl propanoate, propionic acid, butyl ester, propionic acid butyl ester, unii-2nxc4ak99e, butyl propionate natural, fema no. 2211, n-butyl n-propionate
butyl propanoate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapour.

P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c

H226-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

EINECSNumber : 209-669-5

RTECSNumber : UE8245000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only