missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl propionate, 99%

CAS: 590-01-2 | C7H14O2 | 130.187 g/mol

$54.01 - $168.75

Chemical Identifiers

CAS 590-01-2
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00009448
InChI Key BTMVHUNTONAYDX-UHFFFAOYSA-N
Synonym butyl propionate, n-butyl propionate, propanoic acid, butyl ester, n-butyl propanoate, propionic acid, butyl ester, propionic acid butyl ester, unii-2nxc4ak99e, butyl propionate natural, fema no. 2211, n-butyl n-propionate
PubChem CID 11529
IUPAC Name butyl propanoate
SMILES CCCCOC(=O)CC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2012522
View Documents
Thermo Scientific Chemicals
B2012522
100 g
Each for $54.01
Only null left
 
AAB2012536
View Documents
Thermo Scientific Chemicals
B2012536
500 g
Each for $168.75
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

590-01-2
130.187
BTMVHUNTONAYDX-UHFFFAOYSA-N
11529
CCCCOC(=O)CC
C7H14O2
MFCD00009448
butyl propionate, n-butyl propionate, propanoic acid, butyl ester, n-butyl propanoate, propionic acid, butyl ester, propionic acid butyl ester, unii-2nxc4ak99e, butyl propionate natural, fema no. 2211, n-butyl n-propionate
butyl propanoate

Specifications

590-01-2
144°C to 146°C
C7H14O2
MFCD00009448
UN3272
butyl propionate, n-butyl propionate, propanoic acid, butyl ester, n-butyl propanoate, propionic acid, butyl ester, propionic acid butyl ester, unii-2nxc4ak99e, butyl propionate natural, fema no. 2211, n-butyl n-propionate
CCCCOC(=O)CC
130.187
130.19
n-Butyl propionate
0.87
38°C (101°F)
1.402
100 g
14,1587
BTMVHUNTONAYDX-UHFFFAOYSA-N
butyl propanoate
11529
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapour.

P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c

H226-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

EINECSNumber : 209-669-5

RTECSNumber : UE8245000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only