Learn More
n-Butylbenzene, 99+%, Thermo Scientific Chemicals
$135.00 - $329.58
Chemical Identifiers
CAS | 104-51-8 |
---|---|
Molecular Formula | C10H14 |
Molecular Weight (g/mol) | 134.22 |
MDL Number | MFCD00009463 |
InChI Key | OCKPCBLVNKHBMX-UHFFFAOYSA-N |
Synonym | n-butylbenzene, 1-phenylbutane, benzene, butyl, 1-butylbenzene, butyl-benzene, butane, phenyl, unii-s8xz2901rz, butylbenzene, analytical standard, n-butyl benzene, n4b |
PubChem CID | 7705 |
ChEBI | CHEBI:44194 |
IUPAC Name | butylbenzene |
SMILES | CCCCC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC107850500
|
Thermo Scientific Chemicals
107850500 |
50 mL | Glass bottle |
Each for $135.00
|
|
||||
AC107852500
|
Thermo Scientific Chemicals
107852500 |
250 mL | Glass bottle |
Each for $329.58
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
104-51-8 | |
134.22 | |
OCKPCBLVNKHBMX-UHFFFAOYSA-N | |
7705 | |
butylbenzene |
C10H14 | |
MFCD00009463 | |
n-butylbenzene, 1-phenylbutane, benzene, butyl, 1-butylbenzene, butyl-benzene, butane, phenyl, unii-s8xz2901rz, butylbenzene, analytical standard, n-butyl benzene, n4b | |
CHEBI:44194 | |
CCCCC1=CC=CC=C1 |
Specifications
104-51-8 | |
100.0 | |
Colorless | |
183°C | |
Authentic | |
Glass bottle | |
1.4880 to 1.49 | |
MFCD00009463 | |
05,413 | |
15,1551 | |
Solubility in water: insoluble in water. Other solubilities: miscible with ethanol,ether and benzene | |
CCCCC1=CC=CC=C1 | |
134.22 | |
CHEBI:44194 | |
99+% | |
n-Butylbenzene, 99+% |
99.0 | |
-88°C | |
0.8600g/mL | |
59°C | |
99+% | |
C10H14 | |
C6H5(CH2)3CH3 | |
50 mL | |
0.86 | |
n-butylbenzene, 1-phenylbutane, benzene, butyl, 1-butylbenzene, butyl-benzene, butane, phenyl, unii-s8xz2901rz, butylbenzene, analytical standard, n-butyl benzene, n4b | |
OCKPCBLVNKHBMX-UHFFFAOYSA-N | |
butylbenzene | |
7705 | |
134.22 | |
Liquid |
Safety and Handling
GHS H Statement:
Flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 203-209-7
RTECSNumber : CY9070000
TSCA : TSCA
RUO – Research Use Only