missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Butyllithium, 1.6M solution in hexanes, AcroSeal™, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 181275000

 View more versions of this product

Catalog No. AC181275000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

n-Butyllithium
109-72-8,110-54-3
15.25,85.25
Pink to White
60.0°C to 80.0°C
AcroSeal™ Glass Bottle
CH3(CH2)3Li
500 mL
n-butyllithium, lithium, butyl, butyllithium, butyl lithium, n-butyl lithium, lithium butane, n-buli, buli, libu, unii-09w9a6b8zc
[Li]CCCC
64.06
64.06
Powder or Flakes
1.6M solution in hexanes
14.75,84.75
-95.0°C
0.6800g/mL
−18°C
C4H9Li
MFCD00009414
0.68
MZRVEZGGRBJDDB-UHFFFAOYSA-N
lithium;butane
61028
14.0 to 16.0% (as BuLi) (Active base)

Chemical Identifiers

109-72-8
64.06
MZRVEZGGRBJDDB-UHFFFAOYSA-N
61028
C4H9Li
MFCD00009414
n-butyllithium, lithium, butyl, butyllithium, butyl lithium, n-butyl lithium, lithium butane, n-buli, buli, libu, unii-09w9a6b8zc
[Li]CCCC
Videos
Safety and Handling

Safety and Handling

GHS H Statement
In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapour. Catches fire spontaneously if exposed to air. Causes severe skin burns and eye damage. May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure if inhaled. Toxic to aquatic life with long lasting effects. Suspected of damaging fertility. Reacts violently with water.

GHS P Statement
Immediately call a POISON CENTER or doctor/physician. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Avoid release to the environment. Do not breathe dust/fume/gas/mist/vapors/spray. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only