Learn More
n-Butyllithium, 1.6M solution in hexanes, AcroSeal™, Thermo Scientific Chemicals
$125.54 - $295.57
Chemical Identifiers
CAS | 109-72-8 |
---|---|
Molecular Formula | C4H9Li |
Molecular Weight (g/mol) | 64.06 |
MDL Number | MFCD00009414 |
InChI Key | MZRVEZGGRBJDDB-UHFFFAOYSA-N |
Synonym | n-butyllithium, lithium, butyl, butyllithium, butyl lithium, n-butyl lithium, lithium butane, n-buli, buli, libu, unii-09w9a6b8zc |
PubChem CID | 61028 |
SMILES | [Li]CCCC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC181271000
|
Thermo Scientific Chemicals
181271000 |
100 mL | AcroSeal™ Glass bottle |
Each for $125.54
|
|
|||||
AC181275000
|
Thermo Scientific Chemicals
181275000 |
500 mL | AcroSeal™ Glass Bottle |
Each for $257.48
|
|
|||||
AC181278000
|
Thermo Scientific Chemicals
181278000 |
800 mL | AcroSeal™ Glass Bottle |
Each for $295.57
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
109-72-8 | |
64.06 | |
MZRVEZGGRBJDDB-UHFFFAOYSA-N | |
61028 |
C4H9Li | |
MFCD00009414 | |
n-butyllithium, lithium, butyl, butyllithium, butyl lithium, n-butyl lithium, lithium butane, n-buli, buli, libu, unii-09w9a6b8zc | |
[Li]CCCC |
Specifications
109-72-8,110-54-3 | |
15.25,85.25 | |
Pink to White | |
60.0°C to 80.0°C | |
AcroSeal™ Glass bottle | |
CH3(CH2)3Li | |
100 mL | |
n-butyllithium, lithium, butyl, butyllithium, butyl lithium, n-butyl lithium, lithium butane, n-buli, buli, libu, unii-09w9a6b8zc | |
[Li]CCCC | |
64.06 | |
64.06 | |
Powder or Flakes |
14.75,84.75 | |
-95.0°C | |
0.6800g/mL | |
−18°C | |
C4H9Li | |
MFCD00009414 | |
0.68 | |
MZRVEZGGRBJDDB-UHFFFAOYSA-N | |
butyllithium | |
61028 | |
14.0 to 16.0% (as BuLi) (Active base) | |
n-Butyllithium, 1.6M solution in hexanes |
Safety and Handling
GHS H Statement
In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapour. Catches fire spontaneously if exposed to air. Causes severe skin burns and eye damage. May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure if inhaled. Toxic to aquatic life with long lasting effects. Suspected of damaging fertility. Reacts violently with water.
GHS P Statement
Immediately call a POISON CENTER or doctor/physician. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Avoid release to the environment. Do not breathe dust/fume/gas/mist/vapors/spray. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Danger
RUO – Research Use Only