missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Cyclohexylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0356906

 View more versions of this product

Catalog No. AAL0356906


Explore available promotions

Description

Description

N-Cyclohexylhydroxylamine hydrochloride is used in the preparation of 4-carbethoxy-2-cyclohexyl-5-(2H)-isoxazolone and hexahydroisoxazolequinoline analogs. It is also used in the preparation of N-cyclohexyl nitrone by reaction with aldehyde. Further, it acts as a hydroxylamination reagent for allyl esters. In addition to this, it serves as a reagent in the conversion of 1,2-dicarboxylic acids to alfa-beta-unsaturated carboxylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Cyclohexylhydroxylamine hydrochloride is used in the preparation of 4-carbethoxy-2-cyclohexyl-5-(2H)-isoxazolone and hexahydroisoxazolequinoline analogs. It is also used in the preparation of N-cyclohexyl nitrone by reaction with aldehyde. Further, it acts as a hydroxylamination reagent for allyl esters. In addition to this, it serves as a reagent in the conversion of 1,2-dicarboxylic acids to alfa-beta-unsaturated carboxylic acids.

Solubility
Soluble in water.

Notes
Hygroscopic. Moisture sensitive. Incompatible with strong oxidizing agents.
Specifications

Specifications

N-Cyclohexylhydroxylamine hydrochloride
∽166°C (decomposition)
MFCD00012565
3672798
n-cyclohexylhydroxylamine hydrochloride, cyclohexylhydroxylamine hydrochloride, unii-r98vsp3t5l, r98vsp3t5l, cyclohexylhydroxylamine hcl, acmc-1ch2g, n-cyclohexylhydroxylarnine hydrochloride, n-cyclohexyl hydroxylamine hydrochloride, cyclohexanamine,n-hydroxy-,hydrochloride 1:1
SSVAHXZUFFSFER-UHFFFAOYSA-N
N-cyclohexylhydroxylamine;hydrochloride
3084432
98%
25100-12-3
C6H14ClNO
5 g
Hygroscopic
Soluble in water.
C1CCC(CC1)NO.Cl
151.634
151.64

Chemical Identifiers

25100-12-3
151.634
SSVAHXZUFFSFER-UHFFFAOYSA-N
3084432
C1CCC(CC1)NO.Cl
C6H14ClNO
MFCD00012565
n-cyclohexylhydroxylamine hydrochloride, cyclohexylhydroxylamine hydrochloride, unii-r98vsp3t5l, r98vsp3t5l, cyclohexylhydroxylamine hcl, acmc-1ch2g, n-cyclohexylhydroxylarnine hydrochloride, n-cyclohexyl hydroxylamine hydrochloride, cyclohexanamine,n-hydroxy-,hydrochloride 1:1
N-cyclohexylhydroxylamine;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 246-612-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only