Learn More
N-Cyclohexylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
$97.33 - $321.41
Chemical Identifiers
CAS | 25100-12-3 |
---|---|
Molecular Formula | C6H14ClNO |
Molecular Weight (g/mol) | 151.634 |
MDL Number | MFCD00012565 |
InChI Key | SSVAHXZUFFSFER-UHFFFAOYSA-N |
Synonym | n-cyclohexylhydroxylamine hydrochloride, cyclohexylhydroxylamine hydrochloride, unii-r98vsp3t5l, r98vsp3t5l, cyclohexylhydroxylamine hcl, acmc-1ch2g, n-cyclohexylhydroxylarnine hydrochloride, n-cyclohexyl hydroxylamine hydrochloride, cyclohexanamine,n-hydroxy-,hydrochloride 1:1 |
PubChem CID | 3084432 |
IUPAC Name | N-cyclohexylhydroxylamine;hydrochloride |
SMILES | C1CCC(CC1)NO.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL0356903
|
Thermo Scientific Chemicals
L0356903 |
1 g |
Each for $97.33
|
|
||||||
AAL0356906
|
Thermo Scientific Chemicals
L0356906 |
5 g |
Each for $321.41
|
|
||||||
Description
N-Cyclohexylhydroxylamine hydrochloride is used in the preparation of 4-carbethoxy-2-cyclohexyl-5-(2H)-isoxazolone and hexahydroisoxazolequinoline analogs. It is also used in the preparation of N-cyclohexyl nitrone by reaction with aldehyde. Further, it acts as a hydroxylamination reagent for allyl esters. In addition to this, it serves as a reagent in the conversion of 1,2-dicarboxylic acids to alfa-beta-unsaturated carboxylic acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Cyclohexylhydroxylamine hydrochloride is used in the preparation of 4-carbethoxy-2-cyclohexyl-5-(2H)-isoxazolone and hexahydroisoxazolequinoline analogs. It is also used in the preparation of N-cyclohexyl nitrone by reaction with aldehyde. Further, it acts as a hydroxylamination reagent for allyl esters. In addition to this, it serves as a reagent in the conversion of 1,2-dicarboxylic acids to alfa-beta-unsaturated carboxylic acids.
Solubility
Soluble in water.
Notes
Hygroscopic. Moisture sensitive. Incompatible with strong oxidizing agents.
Chemical Identifiers
25100-12-3 | |
151.634 | |
SSVAHXZUFFSFER-UHFFFAOYSA-N | |
3084432 | |
C1CCC(CC1)NO.Cl |
C6H14ClNO | |
MFCD00012565 | |
n-cyclohexylhydroxylamine hydrochloride, cyclohexylhydroxylamine hydrochloride, unii-r98vsp3t5l, r98vsp3t5l, cyclohexylhydroxylamine hcl, acmc-1ch2g, n-cyclohexylhydroxylarnine hydrochloride, n-cyclohexyl hydroxylamine hydrochloride, cyclohexanamine,n-hydroxy-,hydrochloride 1:1 | |
N-cyclohexylhydroxylamine;hydrochloride |
Specifications
25100-12-3 | |
C6H14ClNO | |
1 g | |
Hygroscopic | |
Soluble in water. | |
C1CCC(CC1)NO.Cl | |
151.634 | |
151.64 | |
N-Cyclohexylhydroxylamine hydrochloride |
∽166°C (decomposition) | |
MFCD00012565 | |
3672798 | |
n-cyclohexylhydroxylamine hydrochloride, cyclohexylhydroxylamine hydrochloride, unii-r98vsp3t5l, r98vsp3t5l, cyclohexylhydroxylamine hcl, acmc-1ch2g, n-cyclohexylhydroxylarnine hydrochloride, n-cyclohexyl hydroxylamine hydrochloride, cyclohexanamine,n-hydroxy-,hydrochloride 1:1 | |
SSVAHXZUFFSFER-UHFFFAOYSA-N | |
N-cyclohexylhydroxylamine;hydrochloride | |
3084432 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 246-612-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only