missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Fmoc-L-glutamic acid 1-allyl ester, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6644803

 View more versions of this product

Catalog No. AAH6644803


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

118°C to 122°C
144120-54-7
C23H23NO6
5460531
ORKKMGRINLTBPC-UHFFFAOYNA-N
4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxo-5-(prop-2-en-1-yloxy)pentanoic acid
409.44
409.44
Powder
N-Fmoc-L-glutamic acid 1-allyl ester
Brown
MFCD00467718
fmoc-glu-oall, n-9h-fluorene-9-ylmethoxycarbonyl-l-glutamic acid 1-allyl ester, s-4-9h-fluoren-9-yl methoxy carbonyl amino-5-allyloxy-5-oxopentanoic acid, s-4-9h-fluoren-9-yl methoxy carbonyl amino-5-allyloxy-5-oxopentanoic acid, a-allyl ester, fmoc-glu-allyl ester, fmoc-glu-allyl ester, 1-allyl n-fmoc-l-glutamate, fmoc-glu-oall hplc, 1-allyl n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate, 1-allyl n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate
OC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(=O)OCC=C
1 g
7020606
98%

Chemical Identifiers

144120-54-7
409.44
ORKKMGRINLTBPC-UHFFFAOYNA-N
7020606
OC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(=O)OCC=C
C23H23NO6
MFCD00467718
fmoc-glu-oall, n-9h-fluorene-9-ylmethoxycarbonyl-l-glutamic acid 1-allyl ester, s-4-9h-fluoren-9-yl methoxy carbonyl amino-5-allyloxy-5-oxopentanoic acid, s-4-9h-fluoren-9-yl methoxy carbonyl amino-5-allyloxy-5-oxopentanoic acid, a-allyl ester, fmoc-glu-allyl ester, fmoc-glu-allyl ester, 1-allyl n-fmoc-l-glutamate, fmoc-glu-oall hplc, 1-allyl n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate, 1-allyl n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate
4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxo-5-(prop-2-en-1-yloxy)pentanoic acid
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only