missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Fmoc-L-glutamic acid 5-benzyl ester, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6278306
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
N-Fmoc-L-glutamic acid 5-benzyl ester | |
(decomposition) | |
MFCD00065642 | |
3576722 | |
HJJURMMMGPQIQP-DEOSSOPVSA-N | |
C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 | |
459.498 | |
459.49 |
123639-61-2 | |
C27H25NO6 | |
5 g | |
fmoc-glu obzl-oh, fmoc-l-glutamic acid-gamma-benzyl ester, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-benzyloxy-5-oxopentanoic acid, fmoc-l-glutamic acid 5-benzyl ester, 2s-5-benzyloxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid, pubchem10013, fmoc-glu obzl-oh hplc, n-9-fluorenylmethoxycarbonyl-l-glutamic acid-gamma-benzyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-l-glutamic-acid-gamma-benzyl ester, 2s-2-fluoren-9-ylmethoxy carbonylamino-4-benzyloxycarbonyl butanoic acid | |
+12° (c=1 in Chloroform) | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-phenylmethoxypentanoic acid | |
13966932 | |
95% |
Chemical Identifiers
123639-61-2 | |
459.498 | |
HJJURMMMGPQIQP-DEOSSOPVSA-N | |
13966932 | |
C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 |
C27H25NO6 | |
MFCD00065642 | |
fmoc-glu obzl-oh, fmoc-l-glutamic acid-gamma-benzyl ester, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-benzyloxy-5-oxopentanoic acid, fmoc-l-glutamic acid 5-benzyl ester, 2s-5-benzyloxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid, pubchem10013, fmoc-glu obzl-oh hplc, n-9-fluorenylmethoxycarbonyl-l-glutamic acid-gamma-benzyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-l-glutamic-acid-gamma-benzyl ester, 2s-2-fluoren-9-ylmethoxy carbonylamino-4-benzyloxycarbonyl butanoic acid | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-phenylmethoxypentanoic acid |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold