missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Fmoc-O-tert-butyl-L-allo-threonine, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6657903

 View more versions of this product

Catalog No. AAH6657903


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-Fmoc-O-tert-butyl-L-allo-threonine
White
MFCD00237371
LZOLWEQBVPVDPR-XOBRGWDASA-N
(2S,3S)-3-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
397.47
397.47
Solid
201481-37-0
C23H27NO5
fmoc-allo-thr tbu-oh, fmoc-l-allo-thr tbu-oh, 2s,3s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-tert-butoxy butanoic acid, fmoc-allo-thr tbu, ambotzfaa1655, o-tert-butyl-n-fmoc-l-allothreonine, n-alpha-9-fluorenylmethyloxycarbonyl-o-t-butyl-allo-l-threonine, 2s,3s-2-9h-fluoren-9-ylmethoxycarbonylamino-3-2-methylpropan-2-yl oxy butanoic acid, 2s,3s-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid
C[C@H](OC(C)(C)C)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
1 g
2724634
97%

Chemical Identifiers

201481-37-0
397.47
LZOLWEQBVPVDPR-XOBRGWDASA-N
2724634
C[C@H](OC(C)(C)C)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
C23H27NO5
MFCD00237371
fmoc-allo-thr tbu-oh, fmoc-l-allo-thr tbu-oh, 2s,3s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-tert-butoxy butanoic acid, fmoc-allo-thr tbu, ambotzfaa1655, o-tert-butyl-n-fmoc-l-allothreonine, n-alpha-9-fluorenylmethyloxycarbonyl-o-t-butyl-allo-l-threonine, 2s,3s-2-9h-fluoren-9-ylmethoxycarbonylamino-3-2-methylpropan-2-yl oxy butanoic acid, 2s,3s-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid
(2S,3S)-3-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only