missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Fmoc-O-tert-butyl-L-allo-threonine, 97%, Thermo Scientific Chemicals
$83.36 - $1022.05
Chemical Identifiers
CAS | 201481-37-0 |
---|---|
Molecular Formula | C23H27NO5 |
Molecular Weight (g/mol) | 397.47 |
MDL Number | MFCD00237371 |
InChI Key | LZOLWEQBVPVDPR-XOBRGWDASA-N |
Synonym | fmoc-allo-thr tbu-oh, fmoc-l-allo-thr tbu-oh, 2s,3s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-tert-butoxy butanoic acid, fmoc-allo-thr tbu, ambotzfaa1655, o-tert-butyl-n-fmoc-l-allothreonine, n-alpha-9-fluorenylmethyloxycarbonyl-o-t-butyl-allo-l-threonine, 2s,3s-2-9h-fluoren-9-ylmethoxycarbonylamino-3-2-methylpropan-2-yl oxy butanoic acid, 2s,3s-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid |
PubChem CID | 2724634 |
IUPAC Name | (2S,3S)-3-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid |
SMILES | C[C@H](OC(C)(C)C)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH66579MD
|
Thermo Scientific Chemicals
H66579MD |
250 mg |
Each for $83.36
|
|
|||||
AAH6657903
|
Thermo Scientific Chemicals
H6657903 |
1 g |
Each for $239.96
|
|
|||||
AAH6657906
|
Thermo Scientific Chemicals
H6657906 |
5 g |
Each for $1,022.05
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
201481-37-0 | |
397.47 | |
LZOLWEQBVPVDPR-XOBRGWDASA-N | |
2724634 | |
C[C@H](OC(C)(C)C)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O |
C23H27NO5 | |
MFCD00237371 | |
fmoc-allo-thr tbu-oh, fmoc-l-allo-thr tbu-oh, 2s,3s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-tert-butoxy butanoic acid, fmoc-allo-thr tbu, ambotzfaa1655, o-tert-butyl-n-fmoc-l-allothreonine, n-alpha-9-fluorenylmethyloxycarbonyl-o-t-butyl-allo-l-threonine, 2s,3s-2-9h-fluoren-9-ylmethoxycarbonylamino-3-2-methylpropan-2-yl oxy butanoic acid, 2s,3s-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid | |
(2S,3S)-3-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid |
Specifications
201481-37-0 | |
C23H27NO5 | |
fmoc-allo-thr tbu-oh, fmoc-l-allo-thr tbu-oh, 2s,3s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-tert-butoxy butanoic acid, fmoc-allo-thr tbu, ambotzfaa1655, o-tert-butyl-n-fmoc-l-allothreonine, n-alpha-9-fluorenylmethyloxycarbonyl-o-t-butyl-allo-l-threonine, 2s,3s-2-9h-fluoren-9-ylmethoxycarbonylamino-3-2-methylpropan-2-yl oxy butanoic acid, 2s,3s-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid | |
C[C@H](OC(C)(C)C)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O | |
250 mg | |
2724634 | |
97% | |
N-Fmoc-O-tert-butyl-L-allo-threonine |
White | |
MFCD00237371 | |
LZOLWEQBVPVDPR-XOBRGWDASA-N | |
(2S,3S)-3-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid | |
397.47 | |
397.47 | |
Solid |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only