missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Heptadecane, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 156240250

 View more versions of this product

Catalog No. AC156240250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

n-Heptadecane
22.0°C to 24.0°C
0.7780g/mL
148°C
98.5% min. (GC)
C17H36
CH3(CH2)15CH3
01,173
n-heptadecane, heptadekan, unii-h7c0j39xum, hexadecane, methyl, h7c0j39xum, heptadecane, analytical standard, normal-heptadecane, heptadecane, dsstox_cid_27061, dsstox_rid_82078
NDJKXXJCMXVBJW-UHFFFAOYSA-N
heptadecane
12398
240.47
Liquid After Melting
629-78-7
Colorless
302.0°C
Authentic
Glass bottle
1.4350 to 1.4370
25 mL
0.778
Solubility in water: insoluble. Other solubilities: soluble in alcohol
CCCCCCCCCCCCCCCCC
240.47
CHEBI:16148
99%

Chemical Identifiers

629-78-7
240.47
n-heptadecane, heptadekan, unii-h7c0j39xum, hexadecane, methyl, h7c0j39xum, heptadecane, analytical standard, normal-heptadecane, heptadecane, dsstox_cid_27061, dsstox_rid_82078
CHEBI:16148
CCCCCCCCCCCCCCCCC
C17H36
NDJKXXJCMXVBJW-UHFFFAOYSA-N
12398
heptadecane
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

EINECSNumber : 211-108-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only