missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
n-Heptadecane, 99%, Thermo Scientific Chemicals
$52.95 - $178.53
Chemical Identifiers
CAS | 629-78-7 |
---|---|
Molecular Formula | C17H36 |
Molecular Weight (g/mol) | 240.47 |
InChI Key | NDJKXXJCMXVBJW-UHFFFAOYSA-N |
Synonym | n-heptadecane, heptadekan, unii-h7c0j39xum, hexadecane, methyl, h7c0j39xum, heptadecane, analytical standard, normal-heptadecane, heptadecane, dsstox_cid_27061, dsstox_rid_82078 |
PubChem CID | 12398 |
ChEBI | CHEBI:16148 |
IUPAC Name | heptadecane |
SMILES | CCCCCCCCCCCCCCCCC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC156240250
|
Thermo Scientific Chemicals
156240250 |
25 mL | Glass bottle |
Each for $52.95
|
|
||||
AC156241000
|
Thermo Scientific Chemicals
156241000 |
100 mL | Glass bottle |
Each for $178.53
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
629-78-7 | |
240.47 | |
n-heptadecane, heptadekan, unii-h7c0j39xum, hexadecane, methyl, h7c0j39xum, heptadecane, analytical standard, normal-heptadecane, heptadecane, dsstox_cid_27061, dsstox_rid_82078 | |
CHEBI:16148 | |
CCCCCCCCCCCCCCCCC |
C17H36 | |
NDJKXXJCMXVBJW-UHFFFAOYSA-N | |
12398 | |
heptadecane |
Specifications
629-78-7 | |
Colorless | |
302.0°C | |
Authentic | |
Glass bottle | |
1.4350 to 1.4370 | |
25 mL | |
0.778 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol | |
CCCCCCCCCCCCCCCCC | |
240.47 | |
CHEBI:16148 | |
99% | |
n-Heptadecane |
22.0°C to 24.0°C | |
0.7780g/mL | |
148°C | |
98.5% min. (GC) | |
C17H36 | |
CH3(CH2)15CH3 | |
01,173 | |
n-heptadecane, heptadekan, unii-h7c0j39xum, hexadecane, methyl, h7c0j39xum, heptadecane, analytical standard, normal-heptadecane, heptadecane, dsstox_cid_27061, dsstox_rid_82078 | |
NDJKXXJCMXVBJW-UHFFFAOYSA-N | |
heptadecane | |
12398 | |
240.47 | |
Liquid After Melting |
Safety and Handling
GHS Signal Word: Danger
EINECSNumber : 211-108-4
RUO – Research Use Only