missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methyl-N-[(6-pyrrolidin-1-ylpyridin-2-yl)methyl]amine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC38046DA

 View more versions of this product

Catalog No. CC38046DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

N-Methyl-N-[(6-pyrrolidin-1-ylpyridin-2-yl)methyl]amine
96.5
97%
C11H17N3
1g
GGQYFTXRIMPZQO-UHFFFAOYSA-N
N-methyl-1-(6-pyrrolidin-1-ylpyridin-2-yl)methanamine
24229485
97%
884507-14-6
100.0
Amber Glass Bottle
MFCD09025832
n-methyl-n-6-pyrrolidin-1-ylpyridin-2-yl methyl amine, 2-pyridinemethanamine,n-methyl-6-1-pyrrolidinyl, n-methyl 6-pyrrolidin-1-yl pyridin-2-yl methanamine, methyl 6-pyrrolidin-1-yl pyridin-2-yl methyl amine, methyl 6-pyrrolidinyl 2-pyridyl methyl amine, 2-pyridinemethanamine, n-methyl-6-1-pyrrolidinyl, n-methyl-1-6-1-pyrrolidinyl-2-pyridinyl methanamine, n-methyl-1-6-pyrrolidin-1-yl pyridin-2-yl methanamine
CNCC1=NC(=CC=C1)N2CCCC2
191.278
191.28

Chemical Identifiers

884507-14-6
191.278
GGQYFTXRIMPZQO-UHFFFAOYSA-N
24229485
CNCC1=NC(=CC=C1)N2CCCC2
C11H17N3
MFCD09025832
n-methyl-n-6-pyrrolidin-1-ylpyridin-2-yl methyl amine, 2-pyridinemethanamine,n-methyl-6-1-pyrrolidinyl, n-methyl 6-pyrrolidin-1-yl pyridin-2-yl methanamine, methyl 6-pyrrolidin-1-yl pyridin-2-yl methyl amine, methyl 6-pyrrolidinyl 2-pyridyl methyl amine, 2-pyridinemethanamine, n-methyl-6-1-pyrrolidinyl, n-methyl-1-6-1-pyrrolidinyl-2-pyridinyl methanamine, n-methyl-1-6-pyrrolidin-1-yl pyridin-2-yl methanamine
N-methyl-1-(6-pyrrolidin-1-ylpyridin-2-yl)methanamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions