missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methyl-N-[(6-pyrrolidin-1-ylpyridin-2-yl)methyl]amine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 884507-14-6
Molecular Formula C11H17N3
Molecular Weight (g/mol) 191.278
MDL Number MFCD09025832
InChI Key GGQYFTXRIMPZQO-UHFFFAOYSA-N
Synonym n-methyl-n-6-pyrrolidin-1-ylpyridin-2-yl methyl amine, 2-pyridinemethanamine,n-methyl-6-1-pyrrolidinyl, n-methyl 6-pyrrolidin-1-yl pyridin-2-yl methanamine, methyl 6-pyrrolidin-1-yl pyridin-2-yl methyl amine, methyl 6-pyrrolidinyl 2-pyridyl methyl amine, 2-pyridinemethanamine, n-methyl-6-1-pyrrolidinyl, n-methyl-1-6-1-pyrrolidinyl-2-pyridinyl methanamine, n-methyl-1-6-pyrrolidin-1-yl pyridin-2-yl methanamine
PubChem CID 24229485
IUPAC Name N-methyl-1-(6-pyrrolidin-1-ylpyridin-2-yl)methanamine
SMILES CNCC1=NC(=CC=C1)N2CCCC2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC38046CB
SDS View Documents
Thermo Scientific Chemicals
CC38046CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC38046DA
SDS View Documents
Thermo Scientific Chemicals
CC38046DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

884507-14-6
191.278
GGQYFTXRIMPZQO-UHFFFAOYSA-N
24229485
CNCC1=NC(=CC=C1)N2CCCC2
C11H17N3
MFCD09025832
n-methyl-n-6-pyrrolidin-1-ylpyridin-2-yl methyl amine, 2-pyridinemethanamine,n-methyl-6-1-pyrrolidinyl, n-methyl 6-pyrrolidin-1-yl pyridin-2-yl methanamine, methyl 6-pyrrolidin-1-yl pyridin-2-yl methyl amine, methyl 6-pyrrolidinyl 2-pyridyl methyl amine, 2-pyridinemethanamine, n-methyl-6-1-pyrrolidinyl, n-methyl-1-6-1-pyrrolidinyl-2-pyridinyl methanamine, n-methyl-1-6-pyrrolidin-1-yl pyridin-2-yl methanamine
N-methyl-1-(6-pyrrolidin-1-ylpyridin-2-yl)methanamine

Specifications

884507-14-6
100.0
Amber Glass Bottle
MFCD09025832
n-methyl-n-6-pyrrolidin-1-ylpyridin-2-yl methyl amine, 2-pyridinemethanamine,n-methyl-6-1-pyrrolidinyl, n-methyl 6-pyrrolidin-1-yl pyridin-2-yl methanamine, methyl 6-pyrrolidin-1-yl pyridin-2-yl methyl amine, methyl 6-pyrrolidinyl 2-pyridyl methyl amine, 2-pyridinemethanamine, n-methyl-6-1-pyrrolidinyl, n-methyl-1-6-1-pyrrolidinyl-2-pyridinyl methanamine, n-methyl-1-6-pyrrolidin-1-yl pyridin-2-yl methanamine
CNCC1=NC(=CC=C1)N2CCCC2
191.278
191.28
N-Methyl-N-[(6-pyrrolidin-1-ylpyridin-2-yl)methyl]amine
96.5
97%
C11H17N3
250mg
GGQYFTXRIMPZQO-UHFFFAOYSA-N
N-methyl-1-(6-pyrrolidin-1-ylpyridin-2-yl)methanamine
24229485
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'