missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methylacetamide 99.0+%, TCI America™

Supplier:  TCI America M013325G

 View more versions of this product

Catalog No. M013325G


Explore available promotions

Description

Description

Specifications

Specifications

N-Methylacetamide
White
C3H7NO
25 g
OHLUUHNLEMFGTQ-UHFFFAOYSA-N
N-methylacetamide
6582
73.10
Crystalline Lumps
79-16-3
206°C
MFCD00008683
acetamide, n-methyl, methylacetamide, monomethylacetamide, n-acetyl-n-methylamine, acetylmethylamine, methyl acetamide, n-methyl acetamide, n-methyl-acetamide, acetamide, methyl, n-monomethylacetamide
CC(=O)NC
73.095
CHEBI:87321
≥99.0% (GC)

Chemical Identifiers

79-16-3
73.095
OHLUUHNLEMFGTQ-UHFFFAOYSA-N
6582
N-methylacetamide
C3H7NO
MFCD00008683
acetamide, n-methyl, methylacetamide, monomethylacetamide, n-acetyl-n-methylamine, acetylmethylamine, methyl acetamide, n-methyl acetamide, n-methyl-acetamide, acetamide, methyl, n-monomethylacetamide
CHEBI:87321
CC(=O)NC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3072

RTECSNumber : AC5960000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions