missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Methylacetamide 99.0+%, TCI America™
$31.97 - $70.67
Chemical Identifiers
CAS | 79-16-3 |
---|---|
Molecular Formula | C3H7NO |
Molecular Weight (g/mol) | 73.095 |
MDL Number | MFCD00008683 |
InChI Key | OHLUUHNLEMFGTQ-UHFFFAOYSA-N |
Synonym | acetamide, n-methyl, methylacetamide, monomethylacetamide, n-acetyl-n-methylamine, acetylmethylamine, methyl acetamide, n-methyl acetamide, n-methyl-acetamide, acetamide, methyl, n-monomethylacetamide |
PubChem CID | 6582 |
ChEBI | CHEBI:87321 |
IUPAC Name | N-methylacetamide |
SMILES | CC(=O)NC |
Chemical Identifiers
79-16-3 | |
73.095 | |
OHLUUHNLEMFGTQ-UHFFFAOYSA-N | |
6582 | |
N-methylacetamide |
C3H7NO | |
MFCD00008683 | |
acetamide, n-methyl, methylacetamide, monomethylacetamide, n-acetyl-n-methylamine, acetylmethylamine, methyl acetamide, n-methyl acetamide, n-methyl-acetamide, acetamide, methyl, n-monomethylacetamide | |
CHEBI:87321 | |
CC(=O)NC |
Specifications
79-16-3 | |
206°C | |
MFCD00008683 | |
acetamide, n-methyl, methylacetamide, monomethylacetamide, n-acetyl-n-methylamine, acetylmethylamine, methyl acetamide, n-methyl acetamide, n-methyl-acetamide, acetamide, methyl, n-monomethylacetamide | |
CC(=O)NC | |
73.095 | |
CHEBI:87321 | |
≥99.0% (GC) | |
N-Methylacetamide |
White | |
C3H7NO | |
25 g | |
OHLUUHNLEMFGTQ-UHFFFAOYSA-N | |
N-methylacetamide | |
6582 | |
73.10 | |
Crystalline Lumps |
Safety and Handling
EINECSNumber : (2)-3072
RTECSNumber : AC5960000
TSCA : Yes