missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methylpentylamine 98.0+%, TCI America™

Supplier:  TCI America M22315G

 View more versions of this product

Catalog No. M22315G


Explore available promotions

Description

Description

Specifications

Specifications

N-Methylpentylamine
Yellow
C6H15N
5 g
n-methylpentylamine, n-methylamylamine, amylmethylamine, methyl pentyl amine, 1-pentanamine, n-methyl, methylpentylamine, pentylamine, n-methyl, methyl-pentyl-amine, n-methyl-n-pentylamine, methylamylamine
CCCCCNC
101.19
101.19
Liquid
25419-06-1
118°C
MFCD00041354
2733
UOIWOHLIGKIYFE-UHFFFAOYSA-N
methyl(pentyl)amine
117479
≥98.0% (GC,T)

Chemical Identifiers

25419-06-1
101.19
UOIWOHLIGKIYFE-UHFFFAOYSA-N
117479
CCCCCNC
C6H15N
MFCD00041354
n-methylpentylamine, n-methylamylamine, amylmethylamine, methyl pentyl amine, 1-pentanamine, n-methyl, methylpentylamine, pentylamine, n-methyl, methyl-pentyl-amine, n-methyl-n-pentylamine, methylamylamine
methyl(pentyl)amine
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions