missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Methylpentylamine 98.0+%, TCI America™
$67.30 - $217.06
Chemical Identifiers
CAS | 25419-06-1 |
---|---|
Molecular Formula | C6H15N |
Molecular Weight (g/mol) | 101.19 |
MDL Number | MFCD00041354 |
InChI Key | UOIWOHLIGKIYFE-UHFFFAOYSA-N |
Synonym | n-methylpentylamine, n-methylamylamine, amylmethylamine, methyl pentyl amine, 1-pentanamine, n-methyl, methylpentylamine, pentylamine, n-methyl, methyl-pentyl-amine, n-methyl-n-pentylamine, methylamylamine |
PubChem CID | 117479 |
IUPAC Name | methyl(pentyl)amine |
SMILES | CCCCCNC |
Chemical Identifiers
25419-06-1 | |
101.19 | |
UOIWOHLIGKIYFE-UHFFFAOYSA-N | |
117479 | |
CCCCCNC |
C6H15N | |
MFCD00041354 | |
n-methylpentylamine, n-methylamylamine, amylmethylamine, methyl pentyl amine, 1-pentanamine, n-methyl, methylpentylamine, pentylamine, n-methyl, methyl-pentyl-amine, n-methyl-n-pentylamine, methylamylamine | |
methyl(pentyl)amine |
Specifications
25419-06-1 | |
118°C | |
MFCD00041354 | |
2733 | |
UOIWOHLIGKIYFE-UHFFFAOYSA-N | |
methyl(pentyl)amine | |
117479 | |
≥98.0% (GC,T) | |
N-Methylpentylamine |
Yellow | |
C6H15N | |
1 g | |
n-methylpentylamine, n-methylamylamine, amylmethylamine, methyl pentyl amine, 1-pentanamine, n-methyl, methylpentylamine, pentylamine, n-methyl, methyl-pentyl-amine, n-methyl-n-pentylamine, methylamylamine | |
CCCCCNC | |
101.19 | |
101.19 | |
Liquid |
Safety and Handling
TSCA : No