missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methylpiperidine, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 127480100

 View more versions of this product

Catalog No. AC127480100


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-Methylpiperidine
626-67-5
100.0
Colorless
105.0°C to 108.0°C
Authentic
Glass bottle
1.4368 to 1.4388
10 mL
0.82
Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, acetone, toluene and
[H+].[Cl-].CN1CCCCC1
135.64
5 mPa.s (20°C)
99%
99%
98.5
-50.0°C
0.8200g/mL
3°C
98.5% min. (GC)
C6H14ClN
MFCD00006491
20,16
n-methylpiperidine, piperidine, 1-methyl, methylpiperidine, n-methyl piperidine, 1-methyl-piperidine, n-methylpiperidin, methylpiperdine, 1-methylpiperadine, n-methyl-piperidine, 1-methyl piperidine
QDUXDCXILAPLAG-UHFFFAOYSA-N
1-methylpiperidine
12291
99.18
Liquid

Chemical Identifiers

626-67-5
135.64
QDUXDCXILAPLAG-UHFFFAOYSA-N
12291
[H+].[Cl-].CN1CCCCC1
C6H14ClN
MFCD00006491
n-methylpiperidine, piperidine, 1-methyl, methylpiperidine, n-methyl piperidine, 1-methyl-piperidine, n-methylpiperidin, methylpiperdine, 1-methylpiperadine, n-methyl-piperidine, 1-methyl piperidine
1-methylpiperidine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 210-959-9

RTECSNumber : TN1225000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only