missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methylpiperidine, 99%, Thermo Scientific Chemicals

$55.72 - $167.87

Chemical Identifiers

CAS 626-67-5
Molecular Formula C6H14ClN
Molecular Weight (g/mol) 135.64
MDL Number MFCD00006491
InChI Key QDUXDCXILAPLAG-UHFFFAOYSA-N
Synonym n-methylpiperidine, piperidine, 1-methyl, methylpiperidine, n-methyl piperidine, 1-methyl-piperidine, n-methylpiperidin, methylpiperdine, 1-methylpiperadine, n-methyl-piperidine, 1-methyl piperidine
PubChem CID 12291
IUPAC Name 1-methylpiperidine
SMILES [H+].[Cl-].CN1CCCCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC127480100
SDS View Documents
Thermo Scientific Chemicals
127480100
10 mL Glass bottle
Each for $55.72
 
AC127482500
SDS View Documents
Thermo Scientific Chemicals
127482500
250 mL Glass bottle
Each for $167.87
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

626-67-5
135.64
QDUXDCXILAPLAG-UHFFFAOYSA-N
12291
[H+].[Cl-].CN1CCCCC1
C6H14ClN
MFCD00006491
n-methylpiperidine, piperidine, 1-methyl, methylpiperidine, n-methyl piperidine, 1-methyl-piperidine, n-methylpiperidin, methylpiperdine, 1-methylpiperadine, n-methyl-piperidine, 1-methyl piperidine
1-methylpiperidine

Specifications

626-67-5
100.0
Colorless
105.0°C to 108.0°C
Authentic
Glass bottle
1.4368 to 1.4388
10 mL
0.82
Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, acetone, toluene and
[H+].[Cl-].CN1CCCCC1
135.64
5 mPa.s (20°C)
99%
N-Methylpiperidine, 99%
98.5
-50.0°C
0.8200g/mL
3°C
98.5% min. (GC)
C6H14ClN
MFCD00006491
20,16
n-methylpiperidine, piperidine, 1-methyl, methylpiperidine, n-methyl piperidine, 1-methyl-piperidine, n-methylpiperidin, methylpiperdine, 1-methylpiperadine, n-methyl-piperidine, 1-methyl piperidine
QDUXDCXILAPLAG-UHFFFAOYSA-N
1-methylpiperidine
12291
99.18
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 210-959-9

RTECSNumber : TN1225000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only